Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171869634-171869905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW11
Entrez Gene ID
23291
Gene Name
F-box and WD repeat domain containing 11
Gene Aliases
BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos
UniGene
Hs.484138
Gene Chromosome Location
Chr.5: 171861549-172006873 on Build GRCh38
Transcripts
NM_012300.2 NM_033644.2 NM_033645.2 XM_005265855.4 XM_005265856.2 XM_005265857.2 XM_005265858.2 XM_017009279.1 XM_017009280.1 XM_017009281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5756635 c.1466T>C p.V489A Chr.5: 171869730-171869730 on Build GRCh38
3339518 c.1464G>T p.L488F Chr.5: 171869732-171869732 on Build GRCh38
5819252 c.1374A>G p.Q458Q Chr.5: 171869783-171869783 on Build GRCh38
3702778 c.1389-9C>G p.? Chr.5: 171869816-171869816 on Build GRCh38
4914177 c.1418G>A p.R473H Chr.5: 171869739-171869739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769192430 Chr.5: 171869797-171869797 on Build GRCh38
rs775639820 Chr.5: 171869776-171869776 on Build GRCh38
rs947344716 Chr.5: 171869857-171869857 on Build GRCh38
rs62381942 Chr.5: 171869685-171869685 on Build GRCh38
rs78795140 Chr.5: 171869835-171869835 on Build GRCh38
rs750049684 Chr.5: 171869845-171869845 on Build GRCh38
rs779079729 Chr.5: 171869726-171869726 on Build GRCh38
rs867783413 Chr.5: 171869699-171869699 on Build GRCh38
rs762205049 Chr.5: 171869824-171869824 on Build GRCh38
rs772078839 Chr.5: 171869750-171869750 on Build GRCh38
rs1026298756 Chr.5: 171869802-171869802 on Build GRCh38
rs760981154 Chr.5: 171869710-171869710 on Build GRCh38
rs527971403 Chr.5: 171869709-171869709 on Build GRCh38
rs1017025520 Chr.5: 171869678-171869678 on Build GRCh38
rs777054022 Chr.5: 171869812-171869812 on Build GRCh38
rs747965775 Chr.5: 171869720-171869720 on Build GRCh38
rs773250710 Chr.5: 171869656-171869656 on Build GRCh38
rs773518789 Chr.5: 171869739-171869739 on Build GRCh38
rs751395083 Chr.5: 171869700-171869700 on Build GRCh38
rs17569734 Chr.5: 171869850-171869850 on Build GRCh38
rs974445498 Chr.5: 171869756-171869756 on Build GRCh38
rs769628453 Chr.5: 171869723-171869723 on Build GRCh38
rs752011198 Chr.5: 171869838-171869838 on Build GRCh38
rs143175843 Chr.5: 171869786-171869786 on Build GRCh38

View Full Product Details