Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2705428-2705677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBF4
Entrez Gene ID
57593
Gene Name
early B-cell factor 4
Gene Aliases
COE4, O/E-4
UniGene
Hs.471955
Gene Chromosome Location
Chr.20: 2692878-2760108 on Build GRCh38
Transcripts
NM_001110514.1 XM_006723600.1 XM_017027983.1 XM_017027984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4134381 c.184T>G p.S62A Chr.20: 2705635-2705635 on Build GRCh38
5553543 c.155A>C p.E52A Chr.20: 2705606-2705606 on Build GRCh38
1410907 c.201C>T p.F67F Chr.20: 2705652-2705652 on Build GRCh38
7305773 c.217G>A p.D73N Chr.20: 2705668-2705668 on Build GRCh38
1025444 c.214T>A p.Y72N Chr.20: 2705665-2705665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533417081 Chr.20: 2705485-2705485 on Build GRCh38
rs775299839 Chr.20: 2705647-2705647 on Build GRCh38
rs1029856349 Chr.20: 2705522-2705522 on Build GRCh38
rs368928307 Chr.20: 2705516-2705516 on Build GRCh38
rs1004562888 Chr.20: 2705515-2705515 on Build GRCh38
rs758133887 Chr.20: 2705541-2705541 on Build GRCh38
rs768858190 Chr.20: 2705605-2705605 on Build GRCh38
rs373526286 Chr.20: 2705481-2705481 on Build GRCh38
rs770878009 Chr.20: 2705553-2705553 on Build GRCh38
rs1013654957 Chr.20: 2705534-2705535 on Build GRCh38
rs371167099 Chr.20: 2705547-2705547 on Build GRCh38
rs749562736 Chr.20: 2705512-2705512 on Build GRCh38
rs759588098 Chr.20: 2705557-2705557 on Build GRCh38
rs962689340 Chr.20: 2705601-2705601 on Build GRCh38
rs61729436 Chr.20: 2705620-2705620 on Build GRCh38
rs1040658874 Chr.20: 2705458-2705458 on Build GRCh38
rs951517761 Chr.20: 2705636-2705636 on Build GRCh38
rs533185288 Chr.20: 2705533-2705533 on Build GRCh38
rs762110726 Chr.20: 2705628-2705628 on Build GRCh38
rs566567078 Chr.20: 2705507-2705507 on Build GRCh38
rs1032142772 Chr.20: 2705569-2705569 on Build GRCh38
rs749337026 Chr.20: 2705581-2705581 on Build GRCh38
rs777618354 Chr.20: 2705542-2705542 on Build GRCh38
rs935611050 Chr.20: 2705480-2705480 on Build GRCh38
rs891520361 Chr.20: 2705511-2705511 on Build GRCh38
rs549295636 Chr.20: 2705517-2705517 on Build GRCh38
rs992638167 Chr.20: 2705646-2705646 on Build GRCh38
rs969290317 Chr.20: 2705555-2705555 on Build GRCh38
rs551368995 Chr.20: 2705506-2705506 on Build GRCh38
rs752353320 Chr.20: 2705538-2705538 on Build GRCh38
rs777897169 Chr.20: 2705483-2705483 on Build GRCh38

View Full Product Details