Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59269690-59269951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
UniGene
Hs.607594
Gene Chromosome Location
Chr.10: 59245933-59363186 on Build GRCh38
Transcripts
NM_001001971.2 NM_001143773.1 NM_001166698.1 NM_198215.3 XM_005269618.4 XM_005269619.4 XM_006717702.3 XM_006717703.3 XM_011539490.2 XM_011539491.2 XM_017015885.1 XM_017015886.1 XM_017015887.1 XM_017015888.1 XM_017015889.1 XM_017015890.1 XM_017015891.1 XM_017015892.1 XM_017015893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807402 c.778C>T p.P260S Chr.10: 59269924-59269924 on Build GRCh38
6658597 c.795G>A p.Q265Q Chr.10: 59269907-59269907 on Build GRCh38
3439147 c.771C>T p.A257A Chr.10: 59269931-59269931 on Build GRCh38
88547 c.767C>T p.S256L Chr.10: 59269935-59269935 on Build GRCh38
6658598 c.795G>A p.Q265Q Chr.10: 59269907-59269907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764779870 Chr.10: 59269862-59269862 on Build GRCh38
rs773841608 Chr.10: 59269864-59269864 on Build GRCh38
rs577655249 Chr.10: 59269762-59269762 on Build GRCh38
rs541527042 Chr.10: 59269725-59269725 on Build GRCh38
rs1048992532 Chr.10: 59269819-59269819 on Build GRCh38
rs752371603 Chr.10: 59269886-59269887 on Build GRCh38
rs986293803 Chr.10: 59269775-59269775 on Build GRCh38
rs114253399 Chr.10: 59269814-59269814 on Build GRCh38
rs768712822 Chr.10: 59269783-59269783 on Build GRCh38
rs371828375 Chr.10: 59269791-59269791 on Build GRCh38
rs3217711 Chr.10: 59269725-59269726 on Build GRCh38
rs968172444 Chr.10: 59269774-59269774 on Build GRCh38
rs200777981 Chr.10: 59269897-59269897 on Build GRCh38
rs756416635 Chr.10: 59269901-59269901 on Build GRCh38
rs56080043 Chr.10: 59269735-59269736 on Build GRCh38
rs777854836 Chr.10: 59269907-59269907 on Build GRCh38
rs944712218 Chr.10: 59269806-59269806 on Build GRCh38
rs528359507 Chr.10: 59269813-59269813 on Build GRCh38
rs776959373 Chr.10: 59269790-59269790 on Build GRCh38
rs973526094 Chr.10: 59269759-59269759 on Build GRCh38
rs903106057 Chr.10: 59269817-59269817 on Build GRCh38
rs948527964 Chr.10: 59269904-59269904 on Build GRCh38
rs866421427 Chr.10: 59269740-59269740 on Build GRCh38
rs1022526655 Chr.10: 59269749-59269749 on Build GRCh38
rs374333238 Chr.10: 59269877-59269877 on Build GRCh38
rs752081543 Chr.10: 59269866-59269866 on Build GRCh38
rs1044293423 Chr.10: 59269931-59269931 on Build GRCh38
rs114010355 Chr.10: 59269778-59269778 on Build GRCh38
rs773689302 Chr.10: 59269863-59269863 on Build GRCh38
rs2290852 Chr.10: 59269881-59269881 on Build GRCh38
rs759995374 Chr.10: 59269867-59269867 on Build GRCh38
rs748838575 Chr.10: 59269804-59269804 on Build GRCh38
rs202010856 Chr.10: 59269914-59269914 on Build GRCh38
rs999990758 Chr.10: 59269825-59269825 on Build GRCh38
rs766918029 Chr.10: 59269865-59269865 on Build GRCh38

View Full Product Details