Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138452003-138452246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESYT3
Entrez Gene ID
83850
Gene Name
extended synaptotagmin 3
Gene Aliases
CHR3SYT, E-Syt3, FAM62C
UniGene
Hs.477711
Gene Chromosome Location
Chr.3: 138434573-138479942 on Build GRCh38
Transcripts
NM_001322831.1 NM_001322834.1 NM_031913.4 NR_136409.1 XM_017007307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3587970 c.337C>T p.P113S Chr.3: 138452057-138452057 on Build GRCh38
5398253 c.336C>T p.F112F Chr.3: 138452056-138452056 on Build GRCh38
3587969 c.331C>T p.H111Y Chr.3: 138452051-138452051 on Build GRCh38
5653249 c.349C>T p.R117W Chr.3: 138452069-138452069 on Build GRCh38
4113940 c.329T>G p.I110S Chr.3: 138452049-138452049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771850557 Chr.3: 138452123-138452123 on Build GRCh38
rs752062794 Chr.3: 138452102-138452102 on Build GRCh38
rs375015235 Chr.3: 138452029-138452029 on Build GRCh38
rs758734356 Chr.3: 138452081-138452081 on Build GRCh38
rs1011652208 Chr.3: 138452155-138452155 on Build GRCh38
rs770859872 Chr.3: 138452032-138452032 on Build GRCh38
rs566919461 Chr.3: 138452062-138452062 on Build GRCh38
rs766847982 Chr.3: 138452097-138452097 on Build GRCh38
rs569640885 Chr.3: 138452177-138452177 on Build GRCh38
rs757037174 Chr.3: 138452118-138452118 on Build GRCh38
rs974398146 Chr.3: 138452169-138452169 on Build GRCh38
rs748317354 Chr.3: 138452052-138452053 on Build GRCh38
rs750857861 Chr.3: 138452080-138452080 on Build GRCh38
rs1001609120 Chr.3: 138452030-138452030 on Build GRCh38
rs991970081 Chr.3: 138452069-138452069 on Build GRCh38
rs143292701 Chr.3: 138452051-138452051 on Build GRCh38
rs970207034 Chr.3: 138452188-138452188 on Build GRCh38
rs755582337 Chr.3: 138452107-138452107 on Build GRCh38
rs761880276 Chr.3: 138452138-138452138 on Build GRCh38
rs957672009 Chr.3: 138452147-138452147 on Build GRCh38
rs748878778 Chr.3: 138452028-138452028 on Build GRCh38
rs4678418 Chr.3: 138452200-138452200 on Build GRCh38
rs765436007 Chr.3: 138452076-138452076 on Build GRCh38
rs921662348 Chr.3: 138452182-138452182 on Build GRCh38
rs746900401 Chr.3: 138452132-138452132 on Build GRCh38
rs149154169 Chr.3: 138452049-138452049 on Build GRCh38
rs766189154 Chr.3: 138452075-138452075 on Build GRCh38
rs772174246 Chr.3: 138452050-138452050 on Build GRCh38
rs761311526 Chr.3: 138452049-138452058 on Build GRCh38
rs1023049608 Chr.3: 138452179-138452179 on Build GRCh38
rs769760383 Chr.3: 138452089-138452100 on Build GRCh38
rs377513850 Chr.3: 138452059-138452059 on Build GRCh38
rs535905050 Chr.3: 138452116-138452116 on Build GRCh38
rs371481245 Chr.3: 138452071-138452071 on Build GRCh38
rs811794 Chr.3: 138452119-138452119 on Build GRCh38
rs775321659 Chr.3: 138452124-138452124 on Build GRCh38
rs759769436 Chr.3: 138452074-138452074 on Build GRCh38
rs755598082 Chr.3: 138452026-138452026 on Build GRCh38
rs981630647 Chr.3: 138452195-138452195 on Build GRCh38
rs544372331 Chr.3: 138452193-138452193 on Build GRCh38
rs774344604 Chr.3: 138452033-138452033 on Build GRCh38
rs781371460 Chr.3: 138452039-138452039 on Build GRCh38
rs760693565 Chr.3: 138452058-138452058 on Build GRCh38
rs768632711 Chr.3: 138452135-138452135 on Build GRCh38
rs924936607 Chr.3: 138452143-138452143 on Build GRCh38
rs182078444 Chr.3: 138452120-138452120 on Build GRCh38
rs776831732 Chr.3: 138452136-138452136 on Build GRCh38

View Full Product Details