Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58497050-58497291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP7A1
Entrez Gene ID
1581
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
CP7A, CYP7, CYPVII
UniGene
Hs.1644
Gene Chromosome Location
Chr.8: 58490178-58500161 on Build GRCh38
Transcripts
NM_000780.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4676043 c.339C>T p.S113S Chr.8: 58497173-58497173 on Build GRCh38
1457633 c.451C>T p.R151C Chr.8: 58497061-58497061 on Build GRCh38
3929606 c.378C>T p.N126N Chr.8: 58497134-58497134 on Build GRCh38
3725229 c.397C>A p.L133M Chr.8: 58497115-58497115 on Build GRCh38
6639497 c.357A>G p.G119G Chr.8: 58497155-58497155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763865825 Chr.8: 58497171-58497171 on Build GRCh38
rs759944158 Chr.8: 58497134-58497134 on Build GRCh38
rs780000989 Chr.8: 58497231-58497231 on Build GRCh38
rs757073508 Chr.8: 58497075-58497075 on Build GRCh38
rs781119853 Chr.8: 58497108-58497108 on Build GRCh38
rs752121324 Chr.8: 58497164-58497164 on Build GRCh38
rs1018490874 Chr.8: 58497132-58497132 on Build GRCh38
rs768849156 Chr.8: 58497099-58497099 on Build GRCh38
rs771215892 Chr.8: 58497124-58497124 on Build GRCh38
rs753487730 Chr.8: 58497174-58497174 on Build GRCh38
rs760194463 Chr.8: 58497165-58497165 on Build GRCh38
rs201494047 Chr.8: 58497110-58497110 on Build GRCh38
rs757122872 Chr.8: 58497177-58497177 on Build GRCh38
rs778848131 Chr.8: 58497200-58497200 on Build GRCh38
rs922546527 Chr.8: 58497190-58497190 on Build GRCh38
rs17262728 Chr.8: 58497260-58497260 on Build GRCh38
rs747802295 Chr.8: 58497199-58497199 on Build GRCh38
rs372921624 Chr.8: 58497234-58497234 on Build GRCh38
rs886043760 Chr.8: 58497085-58497085 on Build GRCh38
rs925724879 Chr.8: 58497229-58497229 on Build GRCh38
rs202099580 Chr.8: 58497113-58497113 on Build GRCh38
rs773374797 Chr.8: 58497116-58497116 on Build GRCh38
rs763038606 Chr.8: 58497244-58497244 on Build GRCh38
rs976616207 Chr.8: 58497189-58497189 on Build GRCh38
rs77359018 Chr.8: 58497201-58497201 on Build GRCh38
rs748199397 Chr.8: 58497241-58497241 on Build GRCh38
rs577011584 Chr.8: 58497178-58497178 on Build GRCh38
rs758247511 Chr.8: 58497211-58497211 on Build GRCh38
rs372015675 Chr.8: 58497162-58497162 on Build GRCh38
rs935711242 Chr.8: 58497232-58497232 on Build GRCh38
rs963859142 Chr.8: 58497182-58497182 on Build GRCh38
rs779007978 Chr.8: 58497087-58497087 on Build GRCh38
rs574367449 Chr.8: 58497259-58497259 on Build GRCh38
rs763320209 Chr.8: 58497122-58497122 on Build GRCh38
rs769799586 Chr.8: 58497115-58497115 on Build GRCh38

View Full Product Details