Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123030506-123030779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERL1
Entrez Gene ID
79139
Gene Name
derlin 1
Gene Aliases
DER-1, DER1
UniGene
Hs.241576
Gene Chromosome Location
Chr.8: 123013164-123042423 on Build GRCh38
Transcripts
NM_001134671.2 NM_024295.5 XM_011517299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
461493 c.253C>T p.R85* Chr.8: 123030617-123030617 on Build GRCh38
5450424 c.167T>C p.I56T Chr.8: 123030703-123030703 on Build GRCh38
1644543 c.193G>T p.G65C Chr.8: 123030677-123030677 on Build GRCh38
1454616 c.211C>A p.L71I Chr.8: 123030659-123030659 on Build GRCh38
6352820 c.248C>T p.S83F Chr.8: 123030622-123030622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951471605 Chr.8: 123030574-123030574 on Build GRCh38
rs569638313 Chr.8: 123030723-123030723 on Build GRCh38
rs767277803 Chr.8: 123030753-123030753 on Build GRCh38
rs939962504 Chr.8: 123030573-123030573 on Build GRCh38
rs768374797 Chr.8: 123030734-123030734 on Build GRCh38
rs759161284 Chr.8: 123030609-123030609 on Build GRCh38
rs747509219 Chr.8: 123030711-123030711 on Build GRCh38
rs779341638 Chr.8: 123030565-123030565 on Build GRCh38
rs982777963 Chr.8: 123030576-123030576 on Build GRCh38
rs753793629 Chr.8: 123030677-123030677 on Build GRCh38
rs549756805 Chr.8: 123030675-123030675 on Build GRCh38
rs377046645 Chr.8: 123030602-123030602 on Build GRCh38
rs775066796 Chr.8: 123030715-123030715 on Build GRCh38
rs778478169 Chr.8: 123030701-123030701 on Build GRCh38
rs997442740 Chr.8: 123030581-123030581 on Build GRCh38
rs780179797 Chr.8: 123030676-123030676 on Build GRCh38
rs372443341 Chr.8: 123030721-123030721 on Build GRCh38
rs773833995 Chr.8: 123030738-123030738 on Build GRCh38
rs369499363 Chr.8: 123030748-123030748 on Build GRCh38
rs536205480 Chr.8: 123030616-123030616 on Build GRCh38
rs767472031 Chr.8: 123030642-123030642 on Build GRCh38
rs956879724 Chr.8: 123030696-123030696 on Build GRCh38
rs751380124 Chr.8: 123030636-123030636 on Build GRCh38
rs761546720 Chr.8: 123030640-123030640 on Build GRCh38
rs370493545 Chr.8: 123030608-123030608 on Build GRCh38
rs748405797 Chr.8: 123030594-123030594 on Build GRCh38
rs374946326 Chr.8: 123030635-123030635 on Build GRCh38
rs769491992 Chr.8: 123030712-123030712 on Build GRCh38
rs927168565 Chr.8: 123030690-123030690 on Build GRCh38
rs755243736 Chr.8: 123030695-123030695 on Build GRCh38
rs769410199 Chr.8: 123030612-123030612 on Build GRCh38
rs758808231 Chr.8: 123030699-123030699 on Build GRCh38
rs564730174 Chr.8: 123030617-123030617 on Build GRCh38
rs768970008 Chr.8: 123030560-123030560 on Build GRCh38
rs763585570 Chr.8: 123030618-123030618 on Build GRCh38
rs778955625 Chr.8: 123030698-123030698 on Build GRCh38
rs150123175 Chr.8: 123030557-123030557 on Build GRCh38
rs368249278 Chr.8: 123030651-123030651 on Build GRCh38

View Full Product Details