Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51769055-51769328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS12
Entrez Gene ID
79758
Gene Name
dehydrogenase/reductase 12
Gene Aliases
SDR40C1
UniGene
Hs.266728
Gene Chromosome Location
Chr.13: 51754856-51804206 on Build GRCh38
Transcripts
NM_001270424.1 NM_024705.2 XM_005266527.3 XM_017020750.1 XM_017020751.1 XM_017020754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6001065 c.631G>A p.D211N Chr.13: 51769222-51769222 on Build GRCh38
232158 c.628G>A p.A210T Chr.13: 51769225-51769225 on Build GRCh38
7122622 c.573G>A p.A191A Chr.13: 51769280-51769280 on Build GRCh38
7315118 c.826A>G p.S276G Chr.13: 51769174-51769174 on Build GRCh38
7315034 c.839T>C p.F280S Chr.13: 51769161-51769161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891273638 Chr.13: 51769139-51769139 on Build GRCh38
rs750003918 Chr.13: 51769236-51769236 on Build GRCh38
rs764740741 Chr.13: 51769208-51769209 on Build GRCh38
rs778985501 Chr.13: 51769290-51769290 on Build GRCh38
rs770648455 Chr.13: 51769154-51769154 on Build GRCh38
rs749097745 Chr.13: 51769172-51769172 on Build GRCh38
rs778899689 Chr.13: 51769246-51769246 on Build GRCh38
rs962675943 Chr.13: 51769305-51769305 on Build GRCh38
rs535856517 Chr.13: 51769099-51769099 on Build GRCh38
rs982693064 Chr.13: 51769075-51769075 on Build GRCh38
rs765882230 Chr.13: 51769155-51769155 on Build GRCh38
rs753264454 Chr.13: 51769199-51769199 on Build GRCh38
rs386771009 Chr.13: 51769118-51769119 on Build GRCh38
rs763304893 Chr.13: 51769203-51769203 on Build GRCh38
rs761000580 Chr.13: 51769177-51769177 on Build GRCh38
rs755680235 Chr.13: 51769274-51769274 on Build GRCh38
rs748011826 Chr.13: 51769297-51769297 on Build GRCh38
rs374684388 Chr.13: 51769191-51769191 on Build GRCh38
rs967512488 Chr.13: 51769141-51769141 on Build GRCh38
rs1038041314 Chr.13: 51769104-51769104 on Build GRCh38
rs893393050 Chr.13: 51769184-51769184 on Build GRCh38
rs10676 Chr.13: 51769255-51769255 on Build GRCh38
rs1022980507 Chr.13: 51769200-51769200 on Build GRCh38
rs928786495 Chr.13: 51769162-51769162 on Build GRCh38
rs769925031 Chr.13: 51769259-51769259 on Build GRCh38
rs767126578 Chr.13: 51769232-51769232 on Build GRCh38
rs756952667 Chr.13: 51769231-51769231 on Build GRCh38
rs780037959 Chr.13: 51769168-51769168 on Build GRCh38
rs991698301 Chr.13: 51769166-51769166 on Build GRCh38
rs765720136 Chr.13: 51769192-51769192 on Build GRCh38
rs960554400 Chr.13: 51769073-51769075 on Build GRCh38
rs750092151 Chr.13: 51769272-51769272 on Build GRCh38
rs201005162 Chr.13: 51769174-51769174 on Build GRCh38
rs758279417 Chr.13: 51769301-51769301 on Build GRCh38
rs574218664 Chr.13: 51769275-51769275 on Build GRCh38
rs749278169 Chr.13: 51769241-51769241 on Build GRCh38
rs1050275341 Chr.13: 51769237-51769237 on Build GRCh38
rs767216493 Chr.13: 51769271-51769271 on Build GRCh38
rs764555053 Chr.13: 51769223-51769223 on Build GRCh38
rs763541493 Chr.13: 51769262-51769262 on Build GRCh38
rs775395149 Chr.13: 51769261-51769261 on Build GRCh38
rs774246544 Chr.13: 51769175-51769175 on Build GRCh38
rs138384725 Chr.13: 51769249-51769249 on Build GRCh38
rs960284678 Chr.13: 51769165-51769165 on Build GRCh38
rs145818380 Chr.13: 51769225-51769225 on Build GRCh38
rs143366927 Chr.13: 51769264-51769264 on Build GRCh38
rs754822484 Chr.13: 51769243-51769243 on Build GRCh38
rs920989315 Chr.13: 51769116-51769116 on Build GRCh38
rs779944371 Chr.13: 51769300-51769303 on Build GRCh38
rs953010543 Chr.13: 51769089-51769089 on Build GRCh38
rs375684409 Chr.13: 51769094-51769094 on Build GRCh38
rs199732858 Chr.13: 51769183-51769183 on Build GRCh38
rs1011294178 Chr.13: 51769119-51769119 on Build GRCh38
rs201713739 Chr.13: 51769265-51769265 on Build GRCh38
rs143368917 Chr.13: 51769266-51769266 on Build GRCh38
rs940099103 Chr.13: 51769189-51769189 on Build GRCh38
rs908488619 Chr.13: 51769079-51769079 on Build GRCh38
rs984110717 Chr.13: 51769180-51769180 on Build GRCh38
rs750922810 Chr.13: 51769208-51769208 on Build GRCh38
rs575301482 Chr.13: 51769250-51769250 on Build GRCh38
rs371510695 Chr.13: 51769281-51769281 on Build GRCh38
rs369243854 Chr.13: 51769240-51769240 on Build GRCh38
rs776887240 Chr.13: 51769185-51769185 on Build GRCh38
rs1021403413 Chr.13: 51769136-51769136 on Build GRCh38
rs901639965 Chr.13: 51769215-51769215 on Build GRCh38
rs537934814 Chr.13: 51769248-51769248 on Build GRCh38

View Full Product Details