Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 48758089-48758336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTIF
Entrez Gene ID
9811
Gene Name
cap binding complex dependent translation initiation factor
Gene Aliases
Gm672, KIAA0427
UniGene
Hs.145230
Gene Chromosome Location
Chr.18: 48539056-48863217 on Build GRCh38
Transcripts
NM_001142397.1 NM_014772.2 XM_005258392.4 XM_006722583.3 XM_006722586.3 XM_006722587.3 XM_006722588.3 XM_006722589.3 XM_011526278.2 XM_011526279.2 XM_017026100.1 XM_017026101.1 XM_017026102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3526122 c.770C>T p.P257L Chr.18: 48758248-48758248 on Build GRCh38
4416436 c.637G>A p.D213N Chr.18: 48758115-48758115 on Build GRCh38
6637460 c.785G>A p.R262Q Chr.18: 48758263-48758263 on Build GRCh38
75119 c.799G>A p.A267T Chr.18: 48758133-48758133 on Build GRCh38
5035404 c.883G>T p.E295* Chr.18: 48758217-48758217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775834192 Chr.18: 48758146-48758146 on Build GRCh38
rs773702303 Chr.18: 48758166-48758166 on Build GRCh38
rs933469865 Chr.18: 48758158-48758158 on Build GRCh38
rs143338571 Chr.18: 48758217-48758217 on Build GRCh38
rs754846409 Chr.18: 48758117-48758117 on Build GRCh38
rs767985178 Chr.18: 48758282-48758283 on Build GRCh38
rs778985951 Chr.18: 48758283-48758283 on Build GRCh38
rs771675289 Chr.18: 48758195-48758195 on Build GRCh38
rs752599548 Chr.18: 48758181-48758181 on Build GRCh38
rs755607388 Chr.18: 48758108-48758108 on Build GRCh38
rs1038308 Chr.18: 48758216-48758216 on Build GRCh38
rs534429608 Chr.18: 48758205-48758205 on Build GRCh38
rs762725518 Chr.18: 48758265-48758265 on Build GRCh38
rs772137427 Chr.18: 48758137-48758137 on Build GRCh38
rs369443953 Chr.18: 48758139-48758139 on Build GRCh38
rs549007210 Chr.18: 48758115-48758115 on Build GRCh38
rs762495061 Chr.18: 48758308-48758308 on Build GRCh38
rs375153721 Chr.18: 48758172-48758172 on Build GRCh38
rs559168173 Chr.18: 48758204-48758204 on Build GRCh38
rs748577356 Chr.18: 48758207-48758207 on Build GRCh38
rs377534923 Chr.18: 48758263-48758263 on Build GRCh38
rs777910371 Chr.18: 48758285-48758285 on Build GRCh38
rs758320722 Chr.18: 48758222-48758222 on Build GRCh38
rs767418568 Chr.18: 48758270-48758270 on Build GRCh38
rs200224776 Chr.18: 48758287-48758287 on Build GRCh38
rs764007021 Chr.18: 48758223-48758223 on Build GRCh38
rs144129667 Chr.18: 48758173-48758173 on Build GRCh38
rs759011311 Chr.18: 48758262-48758262 on Build GRCh38
rs781128320 Chr.18: 48758230-48758230 on Build GRCh38
rs1029361999 Chr.18: 48758272-48758272 on Build GRCh38
rs116794939 Chr.18: 48758165-48758165 on Build GRCh38
rs531198993 Chr.18: 48758159-48758159 on Build GRCh38
rs370698446 Chr.18: 48758300-48758300 on Build GRCh38
rs202032754 Chr.18: 48758133-48758133 on Build GRCh38
rs373686942 Chr.18: 48758140-48758140 on Build GRCh38
rs772394794 Chr.18: 48758209-48758209 on Build GRCh38
rs768883954 Chr.18: 48758161-48758161 on Build GRCh38
rs201581371 Chr.18: 48758241-48758241 on Build GRCh38
rs755079998 Chr.18: 48758231-48758231 on Build GRCh38
rs771351730 Chr.18: 48758255-48758255 on Build GRCh38
rs770164138 Chr.18: 48758295-48758295 on Build GRCh38
rs777194047 Chr.18: 48758261-48758261 on Build GRCh38
rs150436238 Chr.18: 48758202-48758202 on Build GRCh38
rs758267207 Chr.18: 48758196-48758196 on Build GRCh38
rs112239580 Chr.18: 48758293-48758293 on Build GRCh38
rs146792603 Chr.18: 48758198-48758198 on Build GRCh38
rs138325558 Chr.18: 48758200-48758200 on Build GRCh38
rs773757307 Chr.18: 48758211-48758211 on Build GRCh38
rs140482288 Chr.18: 48758201-48758201 on Build GRCh38
rs761657690 Chr.18: 48758269-48758269 on Build GRCh38
rs772233727 Chr.18: 48758248-48758248 on Build GRCh38
rs973693680 Chr.18: 48758314-48758314 on Build GRCh38
rs769967502 Chr.18: 48758143-48758143 on Build GRCh38
rs536500179 Chr.18: 48758220-48758220 on Build GRCh38
rs751414020 Chr.18: 48758268-48758268 on Build GRCh38
rs965260656 Chr.18: 48758244-48758244 on Build GRCh38
rs373299394 Chr.18: 48758221-48758221 on Build GRCh38
rs764874698 Chr.18: 48758174-48758174 on Build GRCh38
rs137937120 Chr.18: 48758288-48758288 on Build GRCh38
rs775145496 Chr.18: 48758307-48758307 on Build GRCh38
rs193022476 Chr.18: 48758226-48758226 on Build GRCh38
rs61729840 Chr.18: 48758240-48758240 on Build GRCh38
rs934281397 Chr.18: 48758280-48758280 on Build GRCh38
rs755060893 Chr.18: 48758273-48758273 on Build GRCh38
rs142161540 Chr.18: 48758114-48758114 on Build GRCh38
rs746412292 Chr.18: 48758292-48758292 on Build GRCh38
rs750342612 Chr.18: 48758271-48758271 on Build GRCh38
rs778137644 Chr.18: 48758249-48758249 on Build GRCh38
rs367566894 Chr.18: 48758284-48758284 on Build GRCh38
rs139196062 Chr.18: 48758227-48758227 on Build GRCh38
rs147104990 Chr.18: 48758266-48758266 on Build GRCh38
rs145237824 Chr.18: 48758214-48758214 on Build GRCh38
rs760218099 Chr.18: 48758171-48758171 on Build GRCh38
rs151234284 Chr.18: 48758132-48758132 on Build GRCh38

View Full Product Details