Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 19405689-19405950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSGALNACT1
Entrez Gene ID
55790
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
CSGalNAcT-1, ChGn, beta4GalNAcT
UniGene
Hs.613729
Gene Chromosome Location
Chr.8: 19404161-19757940 on Build GRCh38
Transcripts
NM_001130518.1 NM_018371.4 NR_024040.1 XM_006716358.2 XM_006716359.2 XM_006716360.2 XM_006716361.2 XM_006716362.2 XM_006716363.1 XM_006716364.3 XM_011544578.1 XM_011544579.1 XM_011544580.1 XM_011544582.1 XM_011544583.1 XM_011544584.1 XM_011544585.2 XM_017013623.1 XM_017013624.1 XM_017013625.1 XM_017013626.1 XM_017013627.1 XM_017013628.1 XM_017013629.1 XM_017013630.1 XM_017013631.1 XM_017013632.1 XM_017013633.1 XM_017013634.1 XM_017013635.1 XM_017013636.1 XM_017013637.1 XM_017013638.1 XM_017013639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2784984 c.1526T>G p.M509R Chr.8: 19405853-19405853 on Build GRCh38
2784985 c.1481T>C p.M494T Chr.8: 19405898-19405898 on Build GRCh38
1097804 c.1439G>A p.R480H Chr.8: 19405940-19405940 on Build GRCh38
325070 c.1505C>G p.A502G Chr.8: 19405874-19405874 on Build GRCh38
1097805 c.1438C>T p.R480C Chr.8: 19405941-19405941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758661894 Chr.8: 19405899-19405899 on Build GRCh38
rs763848633 Chr.8: 19405866-19405866 on Build GRCh38
rs563857601 Chr.8: 19405906-19405906 on Build GRCh38
rs767401250 Chr.8: 19405799-19405799 on Build GRCh38
rs146677046 Chr.8: 19405917-19405917 on Build GRCh38
rs758546328 Chr.8: 19405825-19405825 on Build GRCh38
rs777236800 Chr.8: 19405836-19405836 on Build GRCh38
rs762502764 Chr.8: 19405776-19405776 on Build GRCh38
rs752428087 Chr.8: 19405805-19405805 on Build GRCh38
rs751422094 Chr.8: 19405716-19405716 on Build GRCh38
rs780815271 Chr.8: 19405733-19405733 on Build GRCh38
rs930337775 Chr.8: 19405868-19405868 on Build GRCh38
rs779071663 Chr.8: 19405755-19405755 on Build GRCh38
rs61910741 Chr.8: 19405817-19405817 on Build GRCh38
rs572792017 Chr.8: 19405867-19405867 on Build GRCh38
rs148625452 Chr.8: 19405828-19405828 on Build GRCh38
rs140290682 Chr.8: 19405803-19405804 on Build GRCh38
rs756093499 Chr.8: 19405813-19405813 on Build GRCh38
rs376992666 Chr.8: 19405832-19405832 on Build GRCh38
rs896759104 Chr.8: 19405888-19405888 on Build GRCh38
rs1031654903 Chr.8: 19405720-19405720 on Build GRCh38
rs759085046 Chr.8: 19405798-19405798 on Build GRCh38
rs746051125 Chr.8: 19405756-19405756 on Build GRCh38
rs751293282 Chr.8: 19405786-19405786 on Build GRCh38
rs533235539 Chr.8: 19405853-19405853 on Build GRCh38
rs149973638 Chr.8: 19405772-19405772 on Build GRCh38
rs777893668 Chr.8: 19405748-19405748 on Build GRCh38
rs553914582 Chr.8: 19405918-19405918 on Build GRCh38
rs979630298 Chr.8: 19405932-19405932 on Build GRCh38
rs775380173 Chr.8: 19405863-19405863 on Build GRCh38
rs753965541 Chr.8: 19405752-19405753 on Build GRCh38
rs146627040 Chr.8: 19405931-19405931 on Build GRCh38
rs778942732 Chr.8: 19405818-19405818 on Build GRCh38
rs376663557 Chr.8: 19405904-19405904 on Build GRCh38
rs752725076 Chr.8: 19405741-19405741 on Build GRCh38
rs1048728781 Chr.8: 19405869-19405869 on Build GRCh38
rs763973342 Chr.8: 19405815-19405815 on Build GRCh38
rs760350551 Chr.8: 19405865-19405865 on Build GRCh38
rs771167052 Chr.8: 19405753-19405753 on Build GRCh38
rs750404605 Chr.8: 19405891-19405891 on Build GRCh38
rs188875259 Chr.8: 19405879-19405879 on Build GRCh38
rs142112103 Chr.8: 19405861-19405861 on Build GRCh38
rs755074556 Chr.8: 19405767-19405769 on Build GRCh38
rs755989393 Chr.8: 19405747-19405747 on Build GRCh38
rs754762274 Chr.8: 19405726-19405726 on Build GRCh38
rs977533246 Chr.8: 19405719-19405719 on Build GRCh38
rs768980210 Chr.8: 19405763-19405763 on Build GRCh38
rs538811239 Chr.8: 19405745-19405745 on Build GRCh38
rs139076756 Chr.8: 19405929-19405929 on Build GRCh38
rs748633654 Chr.8: 19405837-19405837 on Build GRCh38
rs778908076 Chr.8: 19405784-19405785 on Build GRCh38
rs35971700 Chr.8: 19405930-19405930 on Build GRCh38
rs143957004 Chr.8: 19405826-19405826 on Build GRCh38
rs770610429 Chr.8: 19405839-19405839 on Build GRCh38
rs749311926 Chr.8: 19405749-19405749 on Build GRCh38
rs932909094 Chr.8: 19405822-19405822 on Build GRCh38
rs772537037 Chr.8: 19405758-19405758 on Build GRCh38
rs986904054 Chr.8: 19405831-19405831 on Build GRCh38
rs766161165 Chr.8: 19405777-19405777 on Build GRCh38
rs761314214 Chr.8: 19405762-19405762 on Build GRCh38
rs775705530 Chr.8: 19405759-19405759 on Build GRCh38
rs1036382780 Chr.8: 19405798-19405801 on Build GRCh38
rs757173380 Chr.8: 19405878-19405878 on Build GRCh38
rs372014844 Chr.8: 19405821-19405821 on Build GRCh38
rs944444905 Chr.8: 19405857-19405857 on Build GRCh38
rs1012851863 Chr.8: 19405894-19405894 on Build GRCh38

View Full Product Details