Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10973302-10973549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNND2
Entrez Gene ID
1501
Gene Name
catenin delta 2
Gene Aliases
GT24, NPRAP
UniGene
Hs.314543
Gene Chromosome Location
Chr.5: 10971840-11904067 on Build GRCh38
Transcripts
NM_001288715.1 NM_001288716.1 NM_001288717.1 NM_001332.3 NR_109988.1 XM_005248251.3 XM_005248252.2 XM_005248253.1 XM_011513967.2 XM_017009072.1 XM_017009073.1 XM_017009074.1 XM_017009075.1 XM_017009076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177351 c.3641C>T p.T1214M Chr.5: 10973490-10973490 on Build GRCh38
6637638 c.3663C>T p.P1221P Chr.5: 10973468-10973468 on Build GRCh38
1059439 c.3656C>A p.A1219D Chr.5: 10973475-10973475 on Build GRCh38
2990565 c.3591C>T p.Y1197Y Chr.5: 10973540-10973540 on Build GRCh38
1671507 c.3625G>A p.E1209K Chr.5: 10973506-10973506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018301616 Chr.5: 10973403-10973403 on Build GRCh38
rs149292246 Chr.5: 10973513-10973513 on Build GRCh38
rs377404994 Chr.5: 10973518-10973518 on Build GRCh38
rs751377024 Chr.5: 10973496-10973496 on Build GRCh38
rs750607607 Chr.5: 10973490-10973490 on Build GRCh38
rs1022225005 Chr.5: 10973381-10973381 on Build GRCh38
rs866220973 Chr.5: 10973470-10973470 on Build GRCh38
rs752156033 Chr.5: 10973501-10973501 on Build GRCh38
rs755024571 Chr.5: 10973497-10973497 on Build GRCh38
rs377570490 Chr.5: 10973468-10973468 on Build GRCh38
rs950701492 Chr.5: 10973446-10973446 on Build GRCh38
rs372962432 Chr.5: 10973479-10973479 on Build GRCh38
rs551679968 Chr.5: 10973495-10973495 on Build GRCh38
rs533095262 Chr.5: 10973431-10973431 on Build GRCh38
rs200216751 Chr.5: 10973410-10973410 on Build GRCh38
rs373365622 Chr.5: 10973451-10973451 on Build GRCh38
rs765387726 Chr.5: 10973489-10973489 on Build GRCh38
rs761882768 Chr.5: 10973487-10973487 on Build GRCh38
rs368966775 Chr.5: 10973477-10973477 on Build GRCh38
rs761213897 Chr.5: 10973474-10973474 on Build GRCh38
rs941212509 Chr.5: 10973334-10973334 on Build GRCh38
rs781186044 Chr.5: 10973498-10973498 on Build GRCh38
rs767502731 Chr.5: 10973383-10973383 on Build GRCh38
rs371686103 Chr.5: 10973444-10973444 on Build GRCh38
rs369887146 Chr.5: 10973448-10973448 on Build GRCh38
rs755593916 Chr.5: 10973511-10973511 on Build GRCh38
rs762870714 Chr.5: 10973491-10973491 on Build GRCh38
rs933966444 Chr.5: 10973401-10973401 on Build GRCh38
rs960934519 Chr.5: 10973502-10973502 on Build GRCh38
rs1010452710 Chr.5: 10973375-10973376 on Build GRCh38
rs748902238 Chr.5: 10973517-10973517 on Build GRCh38
rs371937255 Chr.5: 10973432-10973432 on Build GRCh38
rs769401561 Chr.5: 10973441-10973441 on Build GRCh38
rs892044249 Chr.5: 10973344-10973344 on Build GRCh38
rs759466312 Chr.5: 10973341-10973341 on Build GRCh38
rs761840130 Chr.5: 10973418-10973418 on Build GRCh38
rs374759152 Chr.5: 10973437-10973437 on Build GRCh38
rs1006953116 Chr.5: 10973399-10973399 on Build GRCh38
rs192726630 Chr.5: 10973358-10973358 on Build GRCh38

View Full Product Details