Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 208129370-208129643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYGC
Entrez Gene ID
1420
Gene Name
crystallin gamma C
Gene Aliases
CCL, CRYG3, CTRCT2
UniGene
Hs.72910
Gene Chromosome Location
Chr.2: 208128137-208139100 on Build GRCh38
Transcripts
NM_020989.3 XM_011510662.1 NR_038437.1

Gene Symbol
LOC100507443
Entrez Gene ID
100507443
Gene Name
uncharacterized LOC100507443
Gene Aliases
-
UniGene
Hs.570295
Gene Chromosome Location
Chr.2: 208119129-208156762 on Build GRCh38
Transcripts
NM_020989.3 XM_011510662.1 NR_038437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3909431 c.150C>A p.N50K Chr.2: 208129543-208129543 on Build GRCh38
6156877 c.95G>T p.R32L Chr.2: 208129598-208129598 on Build GRCh38
3938890 c.95G>A p.R32H Chr.2: 208129598-208129598 on Build GRCh38
6090372 c.190C>A p.P64T Chr.2: 208129503-208129503 on Build GRCh38
3576942 c.114G>A p.V38V Chr.2: 208129579-208129579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775936147 Chr.2: 208129562-208129562 on Build GRCh38
rs746609116 Chr.2: 208129464-208129464 on Build GRCh38
rs200596535 Chr.2: 208129599-208129599 on Build GRCh38
rs770213432 Chr.2: 208129488-208129488 on Build GRCh38
rs200511472 Chr.2: 208129569-208129569 on Build GRCh38
rs774558801 Chr.2: 208129512-208129512 on Build GRCh38
rs757564058 Chr.2: 208129447-208129447 on Build GRCh38
rs772807273 Chr.2: 208129401-208129401 on Build GRCh38
rs144295934 Chr.2: 208129529-208129529 on Build GRCh38
rs1019289262 Chr.2: 208129460-208129460 on Build GRCh38
rs764296943 Chr.2: 208129427-208129427 on Build GRCh38
rs886902722 Chr.2: 208129576-208129576 on Build GRCh38
rs376249012 Chr.2: 208129414-208129414 on Build GRCh38
rs748143432 Chr.2: 208129462-208129462 on Build GRCh38
rs748198230 Chr.2: 208129398-208129398 on Build GRCh38
rs777748323 Chr.2: 208129541-208129541 on Build GRCh38
rs767906422 Chr.2: 208129453-208129453 on Build GRCh38
rs751761618 Chr.2: 208129445-208129445 on Build GRCh38
rs200203923 Chr.2: 208129404-208129404 on Build GRCh38
rs746192583 Chr.2: 208129477-208129477 on Build GRCh38
rs753295325 Chr.2: 208129425-208129425 on Build GRCh38
rs184571576 Chr.2: 208129573-208129573 on Build GRCh38
rs754845940 Chr.2: 208129455-208129455 on Build GRCh38
rs372299080 Chr.2: 208129514-208129514 on Build GRCh38
rs545258723 Chr.2: 208129456-208129456 on Build GRCh38
rs769985760 Chr.2: 208129556-208129556 on Build GRCh38
rs142890808 Chr.2: 208129609-208129609 on Build GRCh38
rs536312493 Chr.2: 208129619-208129619 on Build GRCh38
rs746520767 Chr.2: 208129544-208129544 on Build GRCh38
rs776052230 Chr.2: 208129607-208129607 on Build GRCh38
rs764211247 Chr.2: 208129503-208129503 on Build GRCh38
rs867018830 Chr.2: 208129451-208129451 on Build GRCh38
rs767818146 Chr.2: 208129515-208129515 on Build GRCh38
rs1040762144 Chr.2: 208129584-208129584 on Build GRCh38
rs749672114 Chr.2: 208129567-208129567 on Build GRCh38
rs369338873 Chr.2: 208129408-208129408 on Build GRCh38
rs61751949 Chr.2: 208129550-208129550 on Build GRCh38
rs765628489 Chr.2: 208129415-208129415 on Build GRCh38
rs1054047596 Chr.2: 208129610-208129610 on Build GRCh38
rs758348597 Chr.2: 208129534-208129534 on Build GRCh38
rs771220059 Chr.2: 208129413-208129413 on Build GRCh38
rs567272897 Chr.2: 208129615-208129615 on Build GRCh38
rs373451856 Chr.2: 208129486-208129486 on Build GRCh38
rs776302681 Chr.2: 208129474-208129474 on Build GRCh38
rs1032571995 Chr.2: 208129476-208129476 on Build GRCh38
rs971216230 Chr.2: 208129526-208129526 on Build GRCh38
rs201699850 Chr.2: 208129463-208129463 on Build GRCh38
rs115828074 Chr.2: 208129570-208129570 on Build GRCh38
rs762892749 Chr.2: 208129426-208129426 on Build GRCh38
rs558534216 Chr.2: 208129500-208129500 on Build GRCh38
rs753573259 Chr.2: 208129454-208129454 on Build GRCh38
rs752526840 Chr.2: 208129596-208129596 on Build GRCh38
rs765112089 Chr.2: 208129525-208129525 on Build GRCh38
rs149880539 Chr.2: 208129517-208129517 on Build GRCh38
rs201193215 Chr.2: 208129397-208129397 on Build GRCh38
rs368378885 Chr.2: 208129410-208129410 on Build GRCh38
rs564915343 Chr.2: 208129598-208129598 on Build GRCh38
rs201568695 Chr.2: 208129551-208129551 on Build GRCh38
rs770559969 Chr.2: 208129473-208129473 on Build GRCh38
rs559463428 Chr.2: 208129572-208129572 on Build GRCh38
rs373774708 Chr.2: 208129549-208129549 on Build GRCh38
rs140859599 Chr.2: 208129583-208129583 on Build GRCh38
rs763520807 Chr.2: 208129501-208129501 on Build GRCh38

View Full Product Details