Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58996516-58996741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSZ
Entrez Gene ID
1522
Gene Name
cathepsin Z
Gene Aliases
CTSX
UniGene
Hs.252549
Gene Chromosome Location
Chr.20: 58995187-59007254 on Build GRCh38
Transcripts
NM_001336.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4581927 c.801+1G>A p.? Chr.20: 58996638-58996638 on Build GRCh38
5565253 c.779G>A p.R260Q Chr.20: 58996661-58996661 on Build GRCh38
5807341 c.750C>T p.I250I Chr.20: 58996690-58996690 on Build GRCh38
1284356 c.797C>A p.P266Q Chr.20: 58996643-58996643 on Build GRCh38
1483790 c.731C>T p.S244F Chr.20: 58996709-58996709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952695862 Chr.20: 58996616-58996616 on Build GRCh38
rs769554603 Chr.20: 58996545-58996546 on Build GRCh38
rs778597258 Chr.20: 58996591-58996591 on Build GRCh38
rs761646955 Chr.20: 58996691-58996691 on Build GRCh38
rs753801317 Chr.20: 58996683-58996683 on Build GRCh38
rs756036366 Chr.20: 58996578-58996578 on Build GRCh38
rs570743873 Chr.20: 58996619-58996619 on Build GRCh38
rs764988308 Chr.20: 58996693-58996693 on Build GRCh38
rs779572666 Chr.20: 58996597-58996597 on Build GRCh38
rs748662783 Chr.20: 58996715-58996715 on Build GRCh38
rs764043042 Chr.20: 58996579-58996579 on Build GRCh38
rs769060257 Chr.20: 58996608-58996608 on Build GRCh38
rs763235073 Chr.20: 58996648-58996648 on Build GRCh38
rs997734091 Chr.20: 58996705-58996705 on Build GRCh38
rs371497156 Chr.20: 58996661-58996661 on Build GRCh38
rs758072520 Chr.20: 58996707-58996707 on Build GRCh38
rs867740289 Chr.20: 58996614-58996614 on Build GRCh38
rs900481339 Chr.20: 58996569-58996569 on Build GRCh38
rs143778696 Chr.20: 58996714-58996714 on Build GRCh38
rs9760 Chr.20: 58996708-58996708 on Build GRCh38
rs773614700 Chr.20: 58996628-58996628 on Build GRCh38
rs537345193 Chr.20: 58996595-58996595 on Build GRCh38
rs113402006 Chr.20: 58996538-58996538 on Build GRCh38
rs558603291 Chr.20: 58996611-58996611 on Build GRCh38
rs759602283 Chr.20: 58996662-58996662 on Build GRCh38
rs770267187 Chr.20: 58996623-58996623 on Build GRCh38
rs549671470 Chr.20: 58996565-58996566 on Build GRCh38
rs764005251 Chr.20: 58996672-58996672 on Build GRCh38
rs199574425 Chr.20: 58996589-58996589 on Build GRCh38
rs773559652 Chr.20: 58996558-58996558 on Build GRCh38
rs746483514 Chr.20: 58996604-58996604 on Build GRCh38
rs762184406 Chr.20: 58996540-58996540 on Build GRCh38
rs904048815 Chr.20: 58996644-58996644 on Build GRCh38
rs750182079 Chr.20: 58996696-58996696 on Build GRCh38
rs763181097 Chr.20: 58996559-58996559 on Build GRCh38
rs892724420 Chr.20: 58996698-58996698 on Build GRCh38
rs752718978 Chr.20: 58996577-58996577 on Build GRCh38
rs771236223 Chr.20: 58996649-58996649 on Build GRCh38
rs753581225 Chr.20: 58996583-58996583 on Build GRCh38
rs751039397 Chr.20: 58996713-58996713 on Build GRCh38
rs79183392 Chr.20: 58996571-58996571 on Build GRCh38
rs762827587 Chr.20: 58996699-58996700 on Build GRCh38
rs745499417 Chr.20: 58996593-58996593 on Build GRCh38
rs765556950 Chr.20: 58996544-58996544 on Build GRCh38

View Full Product Details