Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138062775-138063026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZIP1L
Entrez Gene ID
199221
Gene Name
DAZ interacting zinc finger protein 1 like
Gene Aliases
DZIP2
UniGene
Hs.351403
Gene Chromosome Location
Chr.3: 138061985-138115862 on Build GRCh38
Transcripts
NM_173543.2 XM_005247198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
204920 c.2250G>T p.E750D Chr.3: 138062870-138062870 on Build GRCh38
6891639 c.2178C>T p.S726S Chr.3: 138062942-138062942 on Build GRCh38
3587920 c.2292C>T p.V764V Chr.3: 138062828-138062828 on Build GRCh38
445669 c.2257G>C p.G753R Chr.3: 138062863-138062863 on Build GRCh38
5661604 c.2177C>T p.S726F Chr.3: 138062943-138062943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746953612 Chr.3: 138062833-138062833 on Build GRCh38
rs768188379 Chr.3: 138062884-138062884 on Build GRCh38
rs146029706 Chr.3: 138062887-138062887 on Build GRCh38
rs776036159 Chr.3: 138062886-138062886 on Build GRCh38
rs756771119 Chr.3: 138062798-138062798 on Build GRCh38
rs149714506 Chr.3: 138062926-138062926 on Build GRCh38
rs750050483 Chr.3: 138062823-138062823 on Build GRCh38
rs771430325 Chr.3: 138062867-138062867 on Build GRCh38
rs139991345 Chr.3: 138062906-138062906 on Build GRCh38
rs376534363 Chr.3: 138062995-138062995 on Build GRCh38
rs764752654 Chr.3: 138062896-138062896 on Build GRCh38
rs890924178 Chr.3: 138062846-138062846 on Build GRCh38
rs553034156 Chr.3: 138062916-138062916 on Build GRCh38
rs749469999 Chr.3: 138062824-138062825 on Build GRCh38
rs754972177 Chr.3: 138062835-138062835 on Build GRCh38
rs774693842 Chr.3: 138062993-138062993 on Build GRCh38
rs771337947 Chr.3: 138062983-138062983 on Build GRCh38
rs577055084 Chr.3: 138062883-138062883 on Build GRCh38
rs750055588 Chr.3: 138062900-138062900 on Build GRCh38
rs200913886 Chr.3: 138062832-138062832 on Build GRCh38
rs756163681 Chr.3: 138062934-138062934 on Build GRCh38
rs144562039 Chr.3: 138062930-138062930 on Build GRCh38
rs774927015 Chr.3: 138062874-138062874 on Build GRCh38
rs572225990 Chr.3: 138062928-138062928 on Build GRCh38
rs199601496 Chr.3: 138062847-138062847 on Build GRCh38
rs201921295 Chr.3: 138062997-138062997 on Build GRCh38
rs911042347 Chr.3: 138062859-138062859 on Build GRCh38
rs751410045 Chr.3: 138062908-138062908 on Build GRCh38
rs200902457 Chr.3: 138062820-138062820 on Build GRCh38
rs749378407 Chr.3: 138062963-138062963 on Build GRCh38
rs749471736 Chr.3: 138062852-138062852 on Build GRCh38
rs189060326 Chr.3: 138062938-138062938 on Build GRCh38
rs750918953 Chr.3: 138062982-138062982 on Build GRCh38
rs766015580 Chr.3: 138062907-138062907 on Build GRCh38
rs142617141 Chr.3: 138062836-138062836 on Build GRCh38
rs758211429 Chr.3: 138062829-138062829 on Build GRCh38

View Full Product Details