Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97647243-97647500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX5B
Entrez Gene ID
1329
Gene Name
cytochrome c oxidase subunit 5B
Gene Aliases
COXVB
UniGene
Hs.1342
Gene Chromosome Location
Chr.2: 97646058-97648194 on Build GRCh38
Transcripts
NM_001862.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1632177 c.208_209insCTT p.S71_G72insS Chr.2: 97647374-97647375 on Build GRCh38
2913572 c.188A>G p.N63S Chr.2: 97647354-97647354 on Build GRCh38
5863041 c.195_213del19 p.K68fs*6 Chr.2: 97647361-97647379 on Build GRCh38
6634350 c.191T>C p.V64A Chr.2: 97647357-97647357 on Build GRCh38
6092729 c.217A>T p.T73S Chr.2: 97647383-97647383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765359560 Chr.2: 97647300-97647302 on Build GRCh38
rs753218416 Chr.2: 97647324-97647324 on Build GRCh38
rs779810128 Chr.2: 97647446-97647446 on Build GRCh38
rs768695897 Chr.2: 97647355-97647355 on Build GRCh38
rs768340672 Chr.2: 97647456-97647456 on Build GRCh38
rs983326421 Chr.2: 97647452-97647452 on Build GRCh38
rs751128718 Chr.2: 97647296-97647296 on Build GRCh38
rs777695735 Chr.2: 97647329-97647329 on Build GRCh38
rs75630766 Chr.2: 97647420-97647420 on Build GRCh38
rs749201547 Chr.2: 97647354-97647354 on Build GRCh38
rs902668310 Chr.2: 97647292-97647292 on Build GRCh38
rs753165601 Chr.2: 97647386-97647386 on Build GRCh38
rs775302114 Chr.2: 97647352-97647352 on Build GRCh38
rs991376472 Chr.2: 97647409-97647409 on Build GRCh38
rs756664058 Chr.2: 97647460-97647461 on Build GRCh38
rs952098042 Chr.2: 97647425-97647425 on Build GRCh38
rs773729915 Chr.2: 97647356-97647356 on Build GRCh38
rs959080996 Chr.2: 97647348-97647348 on Build GRCh38
rs886891653 Chr.2: 97647331-97647332 on Build GRCh38
rs763563015 Chr.2: 97647294-97647294 on Build GRCh38
rs377722361 Chr.2: 97647293-97647293 on Build GRCh38
rs747176984 Chr.2: 97647331-97647331 on Build GRCh38
rs752541397 Chr.2: 97647309-97647310 on Build GRCh38
rs772874129 Chr.2: 97647364-97647364 on Build GRCh38
rs761359756 Chr.2: 97647300-97647300 on Build GRCh38
rs756376661 Chr.2: 97647439-97647439 on Build GRCh38
rs1017202545 Chr.2: 97647359-97647359 on Build GRCh38
rs769537358 Chr.2: 97647350-97647350 on Build GRCh38
rs1034743331 Chr.2: 97647325-97647325 on Build GRCh38
rs750051222 Chr.2: 97647305-97647305 on Build GRCh38
rs576346921 Chr.2: 97647328-97647328 on Build GRCh38
rs767273505 Chr.2: 97647306-97647306 on Build GRCh38
rs746108095 Chr.2: 97647340-97647340 on Build GRCh38
rs779446063 Chr.2: 97647314-97647314 on Build GRCh38
rs752017227 Chr.2: 97647400-97647400 on Build GRCh38
rs760438185 Chr.2: 97647378-97647378 on Build GRCh38
rs774288762 Chr.2: 97647461-97647461 on Build GRCh38
rs113948602 Chr.2: 97647342-97647342 on Build GRCh38
rs764591909 Chr.2: 97647396-97647396 on Build GRCh38
rs148368138 Chr.2: 97647392-97647392 on Build GRCh38
rs766952127 Chr.2: 97647358-97647358 on Build GRCh38
rs201514478 Chr.2: 97647305-97647306 on Build GRCh38
rs375680812 Chr.2: 97647450-97647450 on Build GRCh38
rs763699049 Chr.2: 97647320-97647326 on Build GRCh38
rs1042612630 Chr.2: 97647284-97647284 on Build GRCh38
rs753534523 Chr.2: 97647324-97647325 on Build GRCh38
rs1012294517 Chr.2: 97647406-97647406 on Build GRCh38
rs371147857 Chr.2: 97647302-97647302 on Build GRCh38
rs755337358 Chr.2: 97647306-97647306 on Build GRCh38
rs556274805 Chr.2: 97647276-97647276 on Build GRCh38
rs781415911 Chr.2: 97647338-97647338 on Build GRCh38
rs199707194 Chr.2: 97647410-97647410 on Build GRCh38
rs765668278 Chr.2: 97647382-97647382 on Build GRCh38
rs997896123 Chr.2: 97647330-97647330 on Build GRCh38
rs748058788 Chr.2: 97647463-97647463 on Build GRCh38
rs962633438 Chr.2: 97647390-97647390 on Build GRCh38
rs746056555 Chr.2: 97647430-97647430 on Build GRCh38
rs755928818 Chr.2: 97647310-97647314 on Build GRCh38
rs879005992 Chr.2: 97647445-97647446 on Build GRCh38

View Full Product Details