Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170250709-170250970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf58
Entrez Gene ID
133874
Gene Name
chromosome 5 open reading frame 58
Gene Aliases
-
UniGene
Hs.519749
Gene Chromosome Location
Chr.5: 170231730-170252569 on Build GRCh38
Transcripts
NR_131091.1 NR_131092.1 NM_005565.3

Gene Symbol
LCP2
Entrez Gene ID
3937
Gene Name
lymphocyte cytosolic protein 2
Gene Aliases
SLP-76, SLP76
UniGene
Hs.304475
Gene Chromosome Location
Chr.5: 170248084-170297818 on Build GRCh38
Transcripts
NR_131091.1 NR_131092.1 NM_005565.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884193 c.1423C>A p.R475S Chr.5: 170250786-170250786 on Build GRCh38
3614088 c.1373C>T p.P458L Chr.5: 170250836-170250836 on Build GRCh38
6856956 c.1342A>T p.R448* Chr.5: 170250867-170250867 on Build GRCh38
5466387 c.1399G>T p.D467Y Chr.5: 170250810-170250810 on Build GRCh38
1579276 c.1345G>C p.D449H Chr.5: 170250864-170250864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759401598 Chr.5: 170250910-170250910 on Build GRCh38
rs769384862 Chr.5: 170250855-170250855 on Build GRCh38
rs754665101 Chr.5: 170250757-170250757 on Build GRCh38
rs762204104 Chr.5: 170250850-170250850 on Build GRCh38
rs76018609 Chr.5: 170250814-170250814 on Build GRCh38
rs757409371 Chr.5: 170250766-170250766 on Build GRCh38
rs768861788 Chr.5: 170250832-170250832 on Build GRCh38
rs780187792 Chr.5: 170250808-170250808 on Build GRCh38
rs764600817 Chr.5: 170250738-170250738 on Build GRCh38
rs762305943 Chr.5: 170250940-170250940 on Build GRCh38
rs201224616 Chr.5: 170250748-170250748 on Build GRCh38
rs772836771 Chr.5: 170250859-170250859 on Build GRCh38
rs751413816 Chr.5: 170250756-170250756 on Build GRCh38
rs375365359 Chr.5: 170250782-170250782 on Build GRCh38
rs758091686 Chr.5: 170250747-170250747 on Build GRCh38
rs114990719 Chr.5: 170250907-170250907 on Build GRCh38
rs750149893 Chr.5: 170250740-170250740 on Build GRCh38
rs746159185 Chr.5: 170250785-170250785 on Build GRCh38
rs747375090 Chr.5: 170250810-170250810 on Build GRCh38
rs762494899 Chr.5: 170250871-170250871 on Build GRCh38
rs1030709660 Chr.5: 170250780-170250780 on Build GRCh38
rs772264698 Chr.5: 170250796-170250796 on Build GRCh38
rs1036208961 Chr.5: 170250819-170250819 on Build GRCh38
rs368584679 Chr.5: 170250893-170250893 on Build GRCh38
rs777214916 Chr.5: 170250844-170250844 on Build GRCh38

View Full Product Details