Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28696870-28697139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB1
Entrez Gene ID
10691
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
P96PIF, PIF96
UniGene
Hs.632373
Gene Chromosome Location
Chr.1: 28668728-28715603 on Build GRCh38
Transcripts
NM_001319674.1 NM_006582.3 NM_024482.2 XM_011540518.2 XM_011540519.2 XM_011540521.2 XM_011540522.2 XM_017000087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
329373 c.448A>T p.M150L Chr.1: 28696934-28696934 on Build GRCh38
1341599 c.496A>G p.N166D Chr.1: 28696982-28696982 on Build GRCh38
907995 c.544G>T p.A182S Chr.1: 28697030-28697030 on Build GRCh38
6302732 c.580G>A p.V194M Chr.1: 28697036-28697036 on Build GRCh38
6671981 c.463A>G p.I155V Chr.1: 28696949-28696949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764213889 Chr.1: 28697038-28697038 on Build GRCh38
rs778189379 Chr.1: 28696923-28696923 on Build GRCh38
rs755616468 Chr.1: 28696921-28696921 on Build GRCh38
rs751458605 Chr.1: 28697076-28697076 on Build GRCh38
rs772244689 Chr.1: 28696996-28696996 on Build GRCh38
rs766282643 Chr.1: 28697075-28697075 on Build GRCh38
rs775779013 Chr.1: 28697019-28697019 on Build GRCh38
rs375600945 Chr.1: 28697114-28697114 on Build GRCh38
rs140313104 Chr.1: 28697049-28697049 on Build GRCh38
rs146858380 Chr.1: 28697057-28697057 on Build GRCh38
rs370932598 Chr.1: 28697094-28697094 on Build GRCh38
rs760711444 Chr.1: 28697026-28697026 on Build GRCh38
rs116513410 Chr.1: 28696972-28696972 on Build GRCh38
rs935058434 Chr.1: 28697039-28697039 on Build GRCh38
rs746106639 Chr.1: 28696981-28696981 on Build GRCh38
rs200783300 Chr.1: 28697115-28697115 on Build GRCh38
rs145060297 Chr.1: 28696942-28696942 on Build GRCh38
rs767113973 Chr.1: 28697090-28697090 on Build GRCh38
rs747677081 Chr.1: 28696911-28696911 on Build GRCh38
rs754797198 Chr.1: 28697077-28697077 on Build GRCh38
rs749742973 Chr.1: 28696925-28696925 on Build GRCh38

View Full Product Details