Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 1611512-1611751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXC1
Entrez Gene ID
2296
Gene Name
forkhead box C1
Gene Aliases
ARA, FKHL7, FREAC-3, FREAC3, IGDA, IHG1, IRID1, RIEG3
UniGene
Hs.348883
Gene Chromosome Location
Chr.6: 1610446-1613897 on Build GRCh38
Transcripts
NM_001453.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382109 c.1250G>C p.G417A Chr.6: 1611695-1611695 on Build GRCh38
6547052 c.1305G>T p.P435P Chr.6: 1611750-1611750 on Build GRCh38
6382723 c.1249G>C p.G417R Chr.6: 1611694-1611694 on Build GRCh38
398307 c.1077C>T p.S359S Chr.6: 1611522-1611522 on Build GRCh38
1744895 c.1123_1124insGGC p.G375_G376insR Chr.6: 1611568-1611569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749475990 Chr.6: 1611629-1611629 on Build GRCh38
rs949297601 Chr.6: 1611652-1611652 on Build GRCh38
rs1057519482 Chr.6: 1611710-1611710 on Build GRCh38
rs762713984 Chr.6: 1611566-1611604 on Build GRCh38
rs772035500 Chr.6: 1611691-1611691 on Build GRCh38
rs865901577 Chr.6: 1611656-1611656 on Build GRCh38
rs1011576688 Chr.6: 1611723-1611723 on Build GRCh38
rs759491792 Chr.6: 1611690-1611690 on Build GRCh38
rs1020427428 Chr.6: 1611727-1611727 on Build GRCh38
rs922940979 Chr.6: 1611572-1611572 on Build GRCh38
rs398123611 Chr.6: 1611585-1611586 on Build GRCh38
rs986306220 Chr.6: 1611594-1611602 on Build GRCh38
rs796065323 Chr.6: 1611589-1611590 on Build GRCh38
rs748486189 Chr.6: 1611556-1611556 on Build GRCh38
rs760410240 Chr.6: 1611725-1611725 on Build GRCh38
rs767238802 Chr.6: 1611568-1611576 on Build GRCh38
rs572622655 Chr.6: 1611660-1611660 on Build GRCh38
rs975290027 Chr.6: 1611584-1611585 on Build GRCh38
rs908957809 Chr.6: 1611615-1611615 on Build GRCh38
rs751183793 Chr.6: 1611568-1611570 on Build GRCh38
rs730882054 Chr.6: 1611579-1611589 on Build GRCh38
rs545470261 Chr.6: 1611567-1611568 on Build GRCh38
rs1036201687 Chr.6: 1611620-1611620 on Build GRCh38
rs755490120 Chr.6: 1611536-1611536 on Build GRCh38
rs1051933 Chr.6: 1611721-1611721 on Build GRCh38
rs867806300 Chr.6: 1611535-1611535 on Build GRCh38
rs281865460 Chr.6: 1611712-1611712 on Build GRCh38
rs76840944 Chr.6: 1611586-1611587 on Build GRCh38
rs535065221 Chr.6: 1611554-1611554 on Build GRCh38
rs1056318557 Chr.6: 1611698-1611698 on Build GRCh38
rs773512259 Chr.6: 1611618-1611618 on Build GRCh38
rs775432975 Chr.6: 1611720-1611720 on Build GRCh38
rs907221308 Chr.6: 1611683-1611683 on Build GRCh38
rs774351048 Chr.6: 1611657-1611657 on Build GRCh38
rs1001542364 Chr.6: 1611693-1611693 on Build GRCh38
rs770305851 Chr.6: 1611559-1611559 on Build GRCh38
rs1011692878 Chr.6: 1611533-1611533 on Build GRCh38
rs555356900 Chr.6: 1611632-1611632 on Build GRCh38
rs771039137 Chr.6: 1611634-1611634 on Build GRCh38
rs750362304 Chr.6: 1611586-1611600 on Build GRCh38
rs965700446 Chr.6: 1611563-1611601 on Build GRCh38

View Full Product Details