Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200855041-200855314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK1
Entrez Gene ID
1195
Gene Name
CDC like kinase 1
Gene Aliases
CLK, CLK/STY, STY
UniGene
Hs.433732
Gene Chromosome Location
Chr.2: 200853009-200864744 on Build GRCh38
Transcripts
NM_001162407.1 NM_004071.3 NR_027855.1 NR_027856.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5923581 c.1187T>C p.L396P Chr.2: 200855083-200855083 on Build GRCh38
5923582 c.1061T>C p.L354P Chr.2: 200855083-200855083 on Build GRCh38
3990970 c.1203A>C p.P401P Chr.2: 200855067-200855067 on Build GRCh38
5011009 c.1198C>A p.Q400K Chr.2: 200855072-200855072 on Build GRCh38
5011010 c.1072C>A p.Q358K Chr.2: 200855072-200855072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993292442 Chr.2: 200855126-200855126 on Build GRCh38
rs777698575 Chr.2: 200855074-200855074 on Build GRCh38
rs920026212 Chr.2: 200855206-200855206 on Build GRCh38
rs760577178 Chr.2: 200855208-200855208 on Build GRCh38
rs186628206 Chr.2: 200855236-200855236 on Build GRCh38
rs762207952 Chr.2: 200855088-200855089 on Build GRCh38
rs988951230 Chr.2: 200855245-200855245 on Build GRCh38
rs751448751 Chr.2: 200855076-200855076 on Build GRCh38
rs557132334 Chr.2: 200855170-200855170 on Build GRCh38
rs72927093 Chr.2: 200855193-200855193 on Build GRCh38
rs747383341 Chr.2: 200855184-200855188 on Build GRCh38
rs758478473 Chr.2: 200855064-200855064 on Build GRCh38
rs1003026344 Chr.2: 200855285-200855285 on Build GRCh38
rs368071454 Chr.2: 200855120-200855120 on Build GRCh38
rs1023143050 Chr.2: 200855179-200855179 on Build GRCh38
rs370075841 Chr.2: 200855138-200855138 on Build GRCh38
rs914644057 Chr.2: 200855254-200855254 on Build GRCh38
rs768520843 Chr.2: 200855218-200855218 on Build GRCh38
rs571214023 Chr.2: 200855155-200855155 on Build GRCh38
rs1035681776 Chr.2: 200855214-200855214 on Build GRCh38
rs757514465 Chr.2: 200855079-200855079 on Build GRCh38
rs946537687 Chr.2: 200855268-200855268 on Build GRCh38
rs772672605 Chr.2: 200855139-200855143 on Build GRCh38
rs770310485 Chr.2: 200855111-200855112 on Build GRCh38
rs774663559 Chr.2: 200855135-200855135 on Build GRCh38
rs781498072 Chr.2: 200855084-200855084 on Build GRCh38
rs780062775 Chr.2: 200855109-200855109 on Build GRCh38
rs1044478126 Chr.2: 200855228-200855228 on Build GRCh38
rs745876196 Chr.2: 200855085-200855085 on Build GRCh38
rs555126052 Chr.2: 200855237-200855237 on Build GRCh38
rs796607497 Chr.2: 200855169-200855170 on Build GRCh38
rs956116115 Chr.2: 200855223-200855223 on Build GRCh38
rs940036776 Chr.2: 200855190-200855190 on Build GRCh38
rs538747170 Chr.2: 200855164-200855164 on Build GRCh38
rs948781253 Chr.2: 200855215-200855215 on Build GRCh38
rs769919343 Chr.2: 200855090-200855090 on Build GRCh38

View Full Product Details