Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 126996689-126996962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK1
Entrez Gene ID
1793
Gene Name
dedicator of cytokinesis 1
Gene Aliases
DOCK180, ced5
UniGene
Hs.159195
Gene Chromosome Location
Chr.10: 126905409-127452517 on Build GRCh38
Transcripts
NM_001290223.1 NM_001380.4 XM_011539422.2 XM_011539424.1 XM_017015813.1 XM_017015814.1 XM_017015815.1 XM_017015817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122667 c.493G>T p.V165L Chr.10: 126996767-126996767 on Build GRCh38
216822 c.562C>T p.H188Y Chr.10: 126996836-126996836 on Build GRCh38
6647966 c.543G>A p.T181T Chr.10: 126996817-126996817 on Build GRCh38
4842841 c.555C>T p.F185F Chr.10: 126996829-126996829 on Build GRCh38
3978232 c.500G>T p.R167I Chr.10: 126996774-126996774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749105440 Chr.10: 126996784-126996784 on Build GRCh38
rs774339613 Chr.10: 126996810-126996810 on Build GRCh38
rs554024311 Chr.10: 126996829-126996829 on Build GRCh38
rs780629046 Chr.10: 126996778-126996778 on Build GRCh38
rs79043452 Chr.10: 126996817-126996817 on Build GRCh38
rs540730500 Chr.10: 126996876-126996876 on Build GRCh38
rs765575115 Chr.10: 126996823-126996823 on Build GRCh38
rs753140760 Chr.10: 126996764-126996764 on Build GRCh38
rs556176446 Chr.10: 126996739-126996739 on Build GRCh38
rs775277662 Chr.10: 126996717-126996717 on Build GRCh38
rs765627154 Chr.10: 126996763-126996763 on Build GRCh38
rs775853404 Chr.10: 126996901-126996901 on Build GRCh38
rs902631783 Chr.10: 126996752-126996752 on Build GRCh38
rs781331326 Chr.10: 126996894-126996895 on Build GRCh38
rs748051381 Chr.10: 126996892-126996892 on Build GRCh38
rs762809834 Chr.10: 126996726-126996726 on Build GRCh38
rs745471375 Chr.10: 126996779-126996779 on Build GRCh38
rs775644781 Chr.10: 126996860-126996860 on Build GRCh38
rs372867681 Chr.10: 126996913-126996913 on Build GRCh38
rs749004671 Chr.10: 126996875-126996875 on Build GRCh38
rs201203523 Chr.10: 126996762-126996762 on Build GRCh38
rs770984148 Chr.10: 126996816-126996816 on Build GRCh38
rs779660796 Chr.10: 126996873-126996873 on Build GRCh38
rs757412234 Chr.10: 126996742-126996742 on Build GRCh38
rs558950027 Chr.10: 126996890-126996890 on Build GRCh38
rs191930274 Chr.10: 126996871-126996871 on Build GRCh38
rs769567121 Chr.10: 126996716-126996716 on Build GRCh38
rs1014138360 Chr.10: 126996809-126996809 on Build GRCh38
rs372337287 Chr.10: 126996769-126996769 on Build GRCh38
rs761713336 Chr.10: 126996811-126996811 on Build GRCh38
rs749933139 Chr.10: 126996869-126996869 on Build GRCh38
rs34385401 Chr.10: 126996749-126996749 on Build GRCh38
rs754076353 Chr.10: 126996756-126996756 on Build GRCh38
rs751925521 Chr.10: 126996893-126996893 on Build GRCh38
rs756789676 Chr.10: 126996776-126996776 on Build GRCh38
rs757722252 Chr.10: 126996861-126996861 on Build GRCh38
rs752049755 Chr.10: 126996773-126996773 on Build GRCh38
rs779349464 Chr.10: 126996728-126996728 on Build GRCh38
rs768490437 Chr.10: 126996787-126996787 on Build GRCh38
rs895703029 Chr.10: 126996804-126996804 on Build GRCh38
rs752097388 Chr.10: 126996839-126996839 on Build GRCh38
rs538104145 Chr.10: 126996730-126996730 on Build GRCh38
rs764465209 Chr.10: 126996912-126996912 on Build GRCh38
rs770031217 Chr.10: 126996900-126996900 on Build GRCh38
rs762341917 Chr.10: 126996922-126996922 on Build GRCh38
rs746119638 Chr.10: 126996928-126996945 on Build GRCh38
rs563020283 Chr.10: 126996898-126996898 on Build GRCh38
rs763327757 Chr.10: 126996905-126996905 on Build GRCh38
rs758923428 Chr.10: 126996767-126996767 on Build GRCh38
rs769441289 Chr.10: 126996780-126996780 on Build GRCh38
rs768613552 Chr.10: 126996727-126996727 on Build GRCh38
rs766433700 Chr.10: 126996747-126996747 on Build GRCh38
rs577298251 Chr.10: 126996891-126996891 on Build GRCh38
rs779785531 Chr.10: 126996782-126996782 on Build GRCh38
rs764623951 Chr.10: 126996837-126996837 on Build GRCh38
rs367883319 Chr.10: 126996894-126996894 on Build GRCh38
rs751731024 Chr.10: 126996736-126996738 on Build GRCh38
rs769837763 Chr.10: 126996931-126996932 on Build GRCh38
rs759788379 Chr.10: 126996822-126996822 on Build GRCh38
rs755486995 Chr.10: 126996926-126996926 on Build GRCh38
rs780686971 Chr.10: 126996867-126996867 on Build GRCh38

View Full Product Details