Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41391748-41392021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC5
Entrez Gene ID
56915
Gene Name
exosome component 5
Gene Aliases
RRP41B, RRP46, Rrp46p, hRrp46p, p12B
UniGene
Hs.283741
Gene Chromosome Location
Chr.19: 41386370-41397351 on Build GRCh38
Transcripts
NM_020158.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1393966 c.337C>T p.R113C Chr.19: 41391888-41391888 on Build GRCh38
317300 c.345C>A p.S115S Chr.19: 41391880-41391880 on Build GRCh38
3823179 c.308A>T p.E103V Chr.19: 41391917-41391917 on Build GRCh38
5983944 c.328T>C p.L110L Chr.19: 41391897-41391897 on Build GRCh38
997021 c.359T>C p.L120P Chr.19: 41391866-41391866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773082352 Chr.19: 41391793-41391793 on Build GRCh38
rs201990827 Chr.19: 41391846-41391846 on Build GRCh38
rs746157931 Chr.19: 41391850-41391850 on Build GRCh38
rs774110065 Chr.19: 41391931-41391931 on Build GRCh38
rs754815591 Chr.19: 41391815-41391815 on Build GRCh38
rs769484149 Chr.19: 41391860-41391860 on Build GRCh38
rs752998771 Chr.19: 41391822-41391822 on Build GRCh38
rs780526246 Chr.19: 41391802-41391802 on Build GRCh38
rs779618144 Chr.19: 41391847-41391847 on Build GRCh38
rs200307444 Chr.19: 41391898-41391898 on Build GRCh38
rs542429051 Chr.19: 41391884-41391884 on Build GRCh38
rs367849322 Chr.19: 41391888-41391888 on Build GRCh38
rs774277773 Chr.19: 41391874-41391874 on Build GRCh38
rs765428851 Chr.19: 41391988-41391988 on Build GRCh38
rs759499806 Chr.19: 41391934-41391934 on Build GRCh38
rs952451803 Chr.19: 41391852-41391852 on Build GRCh38
rs373660399 Chr.19: 41391919-41391919 on Build GRCh38
rs375717379 Chr.19: 41391938-41391938 on Build GRCh38
rs758907509 Chr.19: 41391991-41391991 on Build GRCh38
rs750516321 Chr.19: 41391989-41391989 on Build GRCh38
rs756710218 Chr.19: 41391797-41391797 on Build GRCh38
rs199563197 Chr.19: 41391945-41391945 on Build GRCh38
rs371719213 Chr.19: 41391828-41391828 on Build GRCh38
rs764481081 Chr.19: 41391893-41391893 on Build GRCh38
rs201243873 Chr.19: 41391857-41391857 on Build GRCh38
rs750970640 Chr.19: 41391909-41391909 on Build GRCh38
rs112271988 Chr.19: 41391873-41391873 on Build GRCh38
rs759515781 Chr.19: 41391767-41391767 on Build GRCh38
rs762854359 Chr.19: 41391794-41391794 on Build GRCh38
rs747858805 Chr.19: 41391859-41391859 on Build GRCh38
rs762509910 Chr.19: 41391872-41391872 on Build GRCh38
rs770459860 Chr.19: 41391924-41391924 on Build GRCh38
rs752487650 Chr.19: 41391887-41391887 on Build GRCh38
rs1025688617 Chr.19: 41391779-41391779 on Build GRCh38
rs780441782 Chr.19: 41391918-41391918 on Build GRCh38
rs138142205 Chr.19: 41391910-41391910 on Build GRCh38
rs906048621 Chr.19: 41391940-41391940 on Build GRCh38
rs772486078 Chr.19: 41391853-41391853 on Build GRCh38
rs767569537 Chr.19: 41391820-41391820 on Build GRCh38
rs772676611 Chr.19: 41391865-41391865 on Build GRCh38
rs759212369 Chr.19: 41391878-41391878 on Build GRCh38
rs200561763 Chr.19: 41391827-41391827 on Build GRCh38
rs868301889 Chr.19: 41391958-41391958 on Build GRCh38
rs775353761 Chr.19: 41391939-41391939 on Build GRCh38
rs878083 Chr.19: 41391783-41391783 on Build GRCh38
rs760601537 Chr.19: 41391941-41391941 on Build GRCh38
rs550529092 Chr.19: 41391913-41391913 on Build GRCh38
rs754115726 Chr.19: 41391967-41391967 on Build GRCh38
rs766371831 Chr.19: 41391807-41391807 on Build GRCh38
rs751344673 Chr.19: 41391810-41391810 on Build GRCh38
rs761941039 Chr.19: 41391980-41391980 on Build GRCh38
rs878082 Chr.19: 41391769-41391769 on Build GRCh38
rs369723905 Chr.19: 41391851-41391851 on Build GRCh38
rs770606591 Chr.19: 41391922-41391922 on Build GRCh38
rs570233269 Chr.19: 41391899-41391899 on Build GRCh38
rs777418116 Chr.19: 41391923-41391923 on Build GRCh38

View Full Product Details