Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152727892-152727635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSAG1
Entrez Gene ID
158511
Gene Name
chondrosarcoma associated gene 1
Gene Aliases
CSAGE, CT24.1
UniGene
Hs.423718
Gene Chromosome Location
Chr.X: 152727484-152733736 on Build GRCh38
Transcripts
NM_001102576.2 NM_153478.2

View Full Product Details