Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32529447-32529720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI1
Entrez Gene ID
51143
Gene Name
dynein cytoplasmic 1 light intermediate chain 1
Gene Aliases
DLC-A, DNCLI1, LIC1
UniGene
Hs.529495
Gene Chromosome Location
Chr.3: 32525971-32570874 on Build GRCh38
Transcripts
NM_016141.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6253694 c.1306+4C>A p.? Chr.3: 32529536-32529536 on Build GRCh38
4679814 c.1288_1289insA p.I430fs*2 Chr.3: 32529557-32529558 on Build GRCh38
4391755 c.1218A>C p.P406P Chr.3: 32529628-32529628 on Build GRCh38
3730878 c.1253C>A p.A418D Chr.3: 32529593-32529593 on Build GRCh38
5014327 c.1225C>G p.P409A Chr.3: 32529621-32529621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373462716 Chr.3: 32529548-32529548 on Build GRCh38
rs374353229 Chr.3: 32529680-32529680 on Build GRCh38
rs35635747 Chr.3: 32529655-32529655 on Build GRCh38
rs748891796 Chr.3: 32529558-32529558 on Build GRCh38
rs770743376 Chr.3: 32529630-32529630 on Build GRCh38
rs779085844 Chr.3: 32529588-32529588 on Build GRCh38
rs368806589 Chr.3: 32529604-32529604 on Build GRCh38
rs775951324 Chr.3: 32529672-32529672 on Build GRCh38
rs779174276 Chr.3: 32529688-32529688 on Build GRCh38
rs376042131 Chr.3: 32529572-32529572 on Build GRCh38
rs1028332339 Chr.3: 32529535-32529535 on Build GRCh38
rs768336349 Chr.3: 32529599-32529599 on Build GRCh38
rs990921555 Chr.3: 32529483-32529483 on Build GRCh38
rs774139319 Chr.3: 32529631-32529631 on Build GRCh38
rs769709439 Chr.3: 32529611-32529611 on Build GRCh38
rs538223495 Chr.3: 32529642-32529642 on Build GRCh38
rs766077643 Chr.3: 32529499-32529499 on Build GRCh38
rs369281136 Chr.3: 32529511-32529511 on Build GRCh38
rs571244328 Chr.3: 32529626-32529626 on Build GRCh38
rs770731383 Chr.3: 32529680-32529681 on Build GRCh38
rs753131248 Chr.3: 32529544-32529544 on Build GRCh38
rs200865606 Chr.3: 32529657-32529657 on Build GRCh38
rs768135842 Chr.3: 32529516-32529516 on Build GRCh38
rs767786234 Chr.3: 32529641-32529641 on Build GRCh38
rs373055308 Chr.3: 32529649-32529649 on Build GRCh38
rs746847924 Chr.3: 32529589-32529589 on Build GRCh38
rs867292994 Chr.3: 32529517-32529517 on Build GRCh38
rs760002247 Chr.3: 32529633-32529633 on Build GRCh38
rs748052246 Chr.3: 32529607-32529607 on Build GRCh38
rs752014900 Chr.3: 32529510-32529510 on Build GRCh38
rs770469657 Chr.3: 32529582-32529582 on Build GRCh38
rs753876334 Chr.3: 32529573-32529573 on Build GRCh38
rs772867764 Chr.3: 32529618-32529618 on Build GRCh38

View Full Product Details