Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 2890663-2890928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMILIN2
Entrez Gene ID
84034
Gene Name
elastin microfibril interfacer 2
Gene Aliases
EMILIN-2, FOAP-10
UniGene
Hs.532815
Gene Chromosome Location
Chr.18: 2847030-2914092 on Build GRCh38
Transcripts
NM_032048.2 XM_017026038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5682432 c.589A>T p.T197S Chr.18: 2890716-2890716 on Build GRCh38
108344 c.612C>T p.S204S Chr.18: 2890739-2890739 on Build GRCh38
5423808 c.611C>A p.S204Y Chr.18: 2890738-2890738 on Build GRCh38
2809118 c.590C>T p.T197M Chr.18: 2890717-2890717 on Build GRCh38
6148916 c.761C>A p.T254N Chr.18: 2890888-2890888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765202061 Chr.18: 2890877-2890877 on Build GRCh38
rs34307586 Chr.18: 2890798-2890798 on Build GRCh38
rs754882796 Chr.18: 2890796-2890796 on Build GRCh38
rs764607642 Chr.18: 2890689-2890691 on Build GRCh38
rs772893330 Chr.18: 2890777-2890777 on Build GRCh38
rs368333068 Chr.18: 2890847-2890847 on Build GRCh38
rs780447143 Chr.18: 2890852-2890852 on Build GRCh38
rs199636328 Chr.18: 2890704-2890704 on Build GRCh38
rs772838075 Chr.18: 2890862-2890862 on Build GRCh38
rs372924807 Chr.18: 2890875-2890875 on Build GRCh38
rs144768923 Chr.18: 2890739-2890739 on Build GRCh38
rs767106127 Chr.18: 2890771-2890771 on Build GRCh38
rs751894221 Chr.18: 2890820-2890820 on Build GRCh38
rs142243285 Chr.18: 2890756-2890756 on Build GRCh38
rs770293374 Chr.18: 2890841-2890841 on Build GRCh38
rs747904549 Chr.18: 2890712-2890712 on Build GRCh38
rs1018674106 Chr.18: 2890715-2890715 on Build GRCh38
rs771172470 Chr.18: 2890728-2890728 on Build GRCh38
rs760175396 Chr.18: 2890779-2890779 on Build GRCh38
rs1034657281 Chr.18: 2890821-2890821 on Build GRCh38
rs36046935 Chr.18: 2890855-2890855 on Build GRCh38
rs766273785 Chr.18: 2890701-2890701 on Build GRCh38
rs1040842224 Chr.18: 2890707-2890707 on Build GRCh38
rs201997785 Chr.18: 2890735-2890735 on Build GRCh38
rs35267664 Chr.18: 2890902-2890902 on Build GRCh38
rs200574377 Chr.18: 2890861-2890861 on Build GRCh38
rs966520860 Chr.18: 2890738-2890738 on Build GRCh38
rs778106576 Chr.18: 2890812-2890812 on Build GRCh38
rs114639659 Chr.18: 2890705-2890705 on Build GRCh38
rs778716795 Chr.18: 2890709-2890709 on Build GRCh38
rs760263461 Chr.18: 2890871-2890871 on Build GRCh38
rs765121960 Chr.18: 2890797-2890797 on Build GRCh38
rs751409842 Chr.18: 2890903-2890903 on Build GRCh38
rs767079167 Chr.18: 2890697-2890697 on Build GRCh38
rs200834200 Chr.18: 2890718-2890718 on Build GRCh38
rs267605169 Chr.18: 2890806-2890806 on Build GRCh38
rs746440776 Chr.18: 2890747-2890747 on Build GRCh38
rs747061429 Chr.18: 2890725-2890725 on Build GRCh38
rs745939565 Chr.18: 2890837-2890837 on Build GRCh38
rs773031433 Chr.18: 2890709-2890710 on Build GRCh38
rs770578467 Chr.18: 2890873-2890873 on Build GRCh38
rs267605170 Chr.18: 2890807-2890807 on Build GRCh38
rs770216845 Chr.18: 2890753-2890753 on Build GRCh38
rs776171192 Chr.18: 2890874-2890874 on Build GRCh38
rs551035165 Chr.18: 2890842-2890842 on Build GRCh38
rs757457887 Chr.18: 2890727-2890738 on Build GRCh38
rs749893470 Chr.18: 2890698-2890698 on Build GRCh38
rs781360194 Chr.18: 2890833-2890833 on Build GRCh38
rs752614087 Chr.18: 2890883-2890883 on Build GRCh38
rs79649705 Chr.18: 2890717-2890717 on Build GRCh38
rs777269499 Chr.18: 2890745-2890745 on Build GRCh38
rs16943977 Chr.18: 2890770-2890770 on Build GRCh38
rs765932028 Chr.18: 2890784-2890784 on Build GRCh38
rs144964001 Chr.18: 2890808-2890808 on Build GRCh38
rs1030172134 Chr.18: 2890726-2890726 on Build GRCh38
rs762834720 Chr.18: 2890901-2890901 on Build GRCh38

View Full Product Details