Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69570450-69570719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRS2
Entrez Gene ID
10818
Gene Name
fibroblast growth factor receptor substrate 2
Gene Aliases
FRS1A, FRS2A, FRS2alpha, SNT, SNT-1, SNT1
UniGene
Hs.593446
Gene Chromosome Location
Chr.12: 69470349-69579793 on Build GRCh38
Transcripts
NM_001042555.2 NM_001278351.1 NM_001278353.1 NM_001278354.1 NM_001278355.1 NM_001278356.1 NM_001278357.1 NM_006654.4 XM_017018717.1 XM_017018718.1 XM_017018719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3384590 c.191G>A p.R64H Chr.12: 69570455-69570455 on Build GRCh38
6451291 c.236G>T p.R79M Chr.12: 69570500-69570500 on Build GRCh38
1606564 c.225A>C p.E75D Chr.12: 69570489-69570489 on Build GRCh38
4210828 c.243A>G p.Q81Q Chr.12: 69570507-69570507 on Build GRCh38
416093 c.233G>A p.R78Q Chr.12: 69570497-69570497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552348406 Chr.12: 69570626-69570626 on Build GRCh38
rs948177502 Chr.12: 69570672-69570672 on Build GRCh38
rs902514095 Chr.12: 69570662-69570662 on Build GRCh38
rs750626746 Chr.12: 69570538-69570538 on Build GRCh38
rs376594597 Chr.12: 69570515-69570515 on Build GRCh38
rs189221320 Chr.12: 69570674-69570674 on Build GRCh38
rs992147850 Chr.12: 69570648-69570648 on Build GRCh38
rs375907210 Chr.12: 69570501-69570501 on Build GRCh38
rs1050282920 Chr.12: 69570599-69570599 on Build GRCh38
rs201673710 Chr.12: 69570537-69570538 on Build GRCh38
rs962192252 Chr.12: 69570610-69570610 on Build GRCh38
rs766577244 Chr.12: 69570470-69570470 on Build GRCh38
rs754263559 Chr.12: 69570569-69570569 on Build GRCh38
rs960663431 Chr.12: 69570647-69570647 on Build GRCh38
rs747696604 Chr.12: 69570529-69570530 on Build GRCh38
rs376616048 Chr.12: 69570634-69570634 on Build GRCh38
rs971966297 Chr.12: 69570614-69570614 on Build GRCh38
rs775249640 Chr.12: 69570684-69570684 on Build GRCh38
rs1042430545 Chr.12: 69570644-69570644 on Build GRCh38
rs879861280 Chr.12: 69570640-69570640 on Build GRCh38
rs12306636 Chr.12: 69570537-69570537 on Build GRCh38
rs527526540 Chr.12: 69570597-69570597 on Build GRCh38
rs147782756 Chr.12: 69570609-69570610 on Build GRCh38
rs1036032954 Chr.12: 69570633-69570633 on Build GRCh38
rs545375580 Chr.12: 69570624-69570629 on Build GRCh38
rs753236534 Chr.12: 69570525-69570525 on Build GRCh38
rs540347888 Chr.12: 69570632-69570644 on Build GRCh38
rs771270808 Chr.12: 69570471-69570471 on Build GRCh38
rs974310139 Chr.12: 69570507-69570507 on Build GRCh38
rs183954168 Chr.12: 69570577-69570577 on Build GRCh38
rs759549467 Chr.12: 69570495-69570495 on Build GRCh38
rs376944090 Chr.12: 69570616-69570617 on Build GRCh38

View Full Product Details