Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81535001-81535240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCN2
Entrez Gene ID
25794
Gene Name
fascin actin-bundling protein 2, retinal
Gene Aliases
RFSN, RP30
UniGene
Hs.118555
Gene Chromosome Location
Chr.17: 81513019-81537130 on Build GRCh38
Transcripts
NM_001077182.2 NM_012418.3 XM_011524587.2 XM_011524590.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5123634 c.966C>T p.H322H Chr.17: 81535191-81535191 on Build GRCh38
1153186 c.867C>T p.H289H Chr.17: 81535092-81535092 on Build GRCh38
5502031 c.834C>T p.N278N Chr.17: 81535059-81535059 on Build GRCh38
4620722 c.935A>G p.Y312C Chr.17: 81535160-81535160 on Build GRCh38
4151798 c.879G>T p.L293L Chr.17: 81535104-81535104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772759235 Chr.17: 81535121-81535121 on Build GRCh38
rs186367879 Chr.17: 81535192-81535192 on Build GRCh38
rs181420326 Chr.17: 81535054-81535054 on Build GRCh38
rs899036308 Chr.17: 81535179-81535179 on Build GRCh38
rs910820925 Chr.17: 81535021-81535021 on Build GRCh38
rs780353525 Chr.17: 81535053-81535053 on Build GRCh38
rs769874646 Chr.17: 81535044-81535044 on Build GRCh38
rs965218161 Chr.17: 81535026-81535026 on Build GRCh38
rs561813825 Chr.17: 81535129-81535129 on Build GRCh38
rs867820179 Chr.17: 81535153-81535153 on Build GRCh38
rs745995346 Chr.17: 81535039-81535039 on Build GRCh38
rs764749597 Chr.17: 81535133-81535133 on Build GRCh38
rs988890666 Chr.17: 81535058-81535058 on Build GRCh38
rs930541410 Chr.17: 81535166-81535166 on Build GRCh38
rs1048953969 Chr.17: 81535168-81535168 on Build GRCh38
rs373043784 Chr.17: 81535093-81535093 on Build GRCh38
rs989667195 Chr.17: 81535036-81535036 on Build GRCh38
rs542647772 Chr.17: 81535123-81535123 on Build GRCh38
rs368687488 Chr.17: 81535092-81535092 on Build GRCh38
rs573355262 Chr.17: 81535218-81535218 on Build GRCh38
rs781235651 Chr.17: 81535035-81535035 on Build GRCh38
rs1037534914 Chr.17: 81535142-81535142 on Build GRCh38
rs376328590 Chr.17: 81535112-81535112 on Build GRCh38
rs575229153 Chr.17: 81535059-81535059 on Build GRCh38
rs927236880 Chr.17: 81535152-81535152 on Build GRCh38
rs757252802 Chr.17: 81535020-81535020 on Build GRCh38
rs367734858 Chr.17: 81535191-81535191 on Build GRCh38

View Full Product Details