Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55624418-55624679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXR2
Entrez Gene ID
139628
Gene Name
forkhead box R2
Gene Aliases
FOXN6
UniGene
Hs.364260
Gene Chromosome Location
Chr.X: 55623400-55626188 on Build GRCh38
Transcripts
NM_198451.3

SNP Detail

SNP ID Chromosome Location
rs753191601 Chr.X: 55624547-55624547 on Build GRCh38
rs973657809 Chr.X: 55624620-55624620 on Build GRCh38
rs771123890 Chr.X: 55624504-55624504 on Build GRCh38
rs769109191 Chr.X: 55624524-55624524 on Build GRCh38
rs1033817883 Chr.X: 55624571-55624571 on Build GRCh38
rs780461913 Chr.X: 55624602-55624602 on Build GRCh38
rs764447916 Chr.X: 55624606-55624606 on Build GRCh38
rs766190976 Chr.X: 55624453-55624453 on Build GRCh38
rs374639616 Chr.X: 55624628-55624628 on Build GRCh38
rs747120288 Chr.X: 55624493-55624493 on Build GRCh38
rs746191989 Chr.X: 55624508-55624508 on Build GRCh38
rs781345083 Chr.X: 55624579-55624579 on Build GRCh38
rs143626914 Chr.X: 55624545-55624545 on Build GRCh38
rs374320995 Chr.X: 55624507-55624507 on Build GRCh38
rs1014381155 Chr.X: 55624516-55624516 on Build GRCh38
rs770167393 Chr.X: 55624514-55624514 on Build GRCh38
rs772763910 Chr.X: 55624534-55624534 on Build GRCh38
rs758941757 Chr.X: 55624474-55624474 on Build GRCh38
rs758347779 Chr.X: 55624479-55624479 on Build GRCh38
rs752505595 Chr.X: 55624546-55624546 on Build GRCh38
rs148892756 Chr.X: 55624544-55624544 on Build GRCh38
rs745397944 Chr.X: 55624623-55624623 on Build GRCh38
rs760370199 Chr.X: 55624451-55624451 on Build GRCh38
rs144748891 Chr.X: 55624489-55624489 on Build GRCh38
rs752698583 Chr.X: 55624478-55624478 on Build GRCh38
rs138589958 Chr.X: 55624543-55624543 on Build GRCh38
rs371034151 Chr.X: 55624473-55624473 on Build GRCh38
rs1032947987 Chr.X: 55624611-55624611 on Build GRCh38
rs746103591 Chr.X: 55624599-55624599 on Build GRCh38
rs778422695 Chr.X: 55624587-55624587 on Build GRCh38
rs757360630 Chr.X: 55624569-55624569 on Build GRCh38
rs753662592 Chr.X: 55624467-55624467 on Build GRCh38
rs756014275 Chr.X: 55624468-55624468 on Build GRCh38
rs757548761 Chr.X: 55624629-55624629 on Build GRCh38
rs764641655 Chr.X: 55624620-55624622 on Build GRCh38
rs760150518 Chr.X: 55624539-55624539 on Build GRCh38
rs755548092 Chr.X: 55624535-55624536 on Build GRCh38
rs754858028 Chr.X: 55624472-55624472 on Build GRCh38
rs2375465 Chr.X: 55624568-55624568 on Build GRCh38
rs375881019 Chr.X: 55624565-55624565 on Build GRCh38
rs372713124 Chr.X: 55624515-55624515 on Build GRCh38
rs372245146 Chr.X: 55624618-55624618 on Build GRCh38
rs181089633 Chr.X: 55624540-55624540 on Build GRCh38

View Full Product Details