Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 101979724-101979959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1H1
Entrez Gene ID
1778
Gene Name
dynein cytoplasmic 1 heavy chain 1
Gene Aliases
CMT2O, DHC1, DHC1a, DNCH1, DNCL, DNECL, DYHC, Dnchc1, HL-3, SMALED1, p22
UniGene
Hs.614080
Gene Chromosome Location
Chr.14: 101964528-102050798 on Build GRCh38
Transcripts
NM_001376.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7181027 c.726T>C p.L242L Chr.14: 101979926-101979926 on Build GRCh38
140050 c.718A>T p.T240S Chr.14: 101979918-101979918 on Build GRCh38
3744216 c.680C>T p.P227L Chr.14: 101979880-101979880 on Build GRCh38
4803224 c.667C>G p.R223G Chr.14: 101979867-101979867 on Build GRCh38
2262120 c.667C>T p.R223C Chr.14: 101979867-101979867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037520835 Chr.14: 101979896-101979896 on Build GRCh38
rs769311346 Chr.14: 101979902-101979902 on Build GRCh38
rs144882836 Chr.14: 101979785-101979785 on Build GRCh38
rs561761105 Chr.14: 101979868-101979868 on Build GRCh38
rs200918195 Chr.14: 101979767-101979767 on Build GRCh38
rs138702876 Chr.14: 101979809-101979809 on Build GRCh38
rs758747746 Chr.14: 101979825-101979825 on Build GRCh38
rs556010695 Chr.14: 101979823-101979823 on Build GRCh38
rs995994279 Chr.14: 101979907-101979907 on Build GRCh38
rs745425454 Chr.14: 101979879-101979879 on Build GRCh38
rs77216005 Chr.14: 101979796-101979796 on Build GRCh38
rs941435600 Chr.14: 101979893-101979893 on Build GRCh38
rs3818188 Chr.14: 101979824-101979824 on Build GRCh38
rs757819428 Chr.14: 101979867-101979867 on Build GRCh38
rs761850867 Chr.14: 101979764-101979764 on Build GRCh38
rs768502998 Chr.14: 101979918-101979918 on Build GRCh38
rs768596689 Chr.14: 101979752-101979752 on Build GRCh38
rs376456164 Chr.14: 101979906-101979906 on Build GRCh38
rs748927251 Chr.14: 101979914-101979914 on Build GRCh38
rs992852984 Chr.14: 101979807-101979807 on Build GRCh38
rs774395446 Chr.14: 101979761-101979761 on Build GRCh38
rs371221480 Chr.14: 101979919-101979919 on Build GRCh38
rs138182529 Chr.14: 101979834-101979834 on Build GRCh38
rs374440380 Chr.14: 101979857-101979857 on Build GRCh38

View Full Product Details