Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117824557-117824786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD2
Entrez Gene ID
486
Gene Name
FXYD domain containing ion transport regulator 2
Gene Aliases
ATP1G1, HOMG2
UniGene
Hs.731865
Gene Chromosome Location
Chr.11: 117820075-117828092 on Build GRCh38
Transcripts
NM_001680.4 NM_021603.3 NM_001204268.1 NM_001243598.2

Gene Symbol
FXYD6-FXYD2
Entrez Gene ID
100533181
Gene Name
FXYD6-FXYD2 readthrough
Gene Aliases
-
UniGene
Hs.744850
Gene Chromosome Location
Chr.11: 117820075-117877031 on Build GRCh38
Transcripts
NM_001680.4 NM_021603.3 NM_001204268.1 NM_001243598.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7250718 c.366T>C p.H122H Chr.11: 117824663-117824663 on Build GRCh38
1704596 c.363C>T p.V121V Chr.11: 117824660-117824660 on Build GRCh38
5837893 c.368G>A p.R123Q Chr.11: 117824665-117824665 on Build GRCh38
6667978 c.361G>A p.V121I Chr.11: 117824658-117824658 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762618472 Chr.11: 117824700-117824700 on Build GRCh38
rs902662717 Chr.11: 117824660-117824660 on Build GRCh38
rs377559962 Chr.11: 117824635-117824635 on Build GRCh38
rs972474595 Chr.11: 117824624-117824624 on Build GRCh38
rs561338739 Chr.11: 117824705-117824705 on Build GRCh38
rs754455934 Chr.11: 117824639-117824639 on Build GRCh38
rs760168417 Chr.11: 117824687-117824687 on Build GRCh38
rs752142826 Chr.11: 117824727-117824727 on Build GRCh38
rs780734647 Chr.11: 117824644-117824644 on Build GRCh38
rs866309547 Chr.11: 117824734-117824734 on Build GRCh38
rs909741310 Chr.11: 117824593-117824593 on Build GRCh38
rs372484079 Chr.11: 117824629-117824629 on Build GRCh38
rs373105708 Chr.11: 117824632-117824632 on Build GRCh38
rs775779800 Chr.11: 117824689-117824689 on Build GRCh38
rs764165453 Chr.11: 117824693-117824693 on Build GRCh38
rs1055577778 Chr.11: 117824621-117824621 on Build GRCh38
rs371012665 Chr.11: 117824658-117824658 on Build GRCh38
rs200896094 Chr.11: 117824696-117824696 on Build GRCh38
rs750118630 Chr.11: 117824633-117824633 on Build GRCh38
rs771733437 Chr.11: 117824656-117824656 on Build GRCh38
rs746807217 Chr.11: 117824664-117824664 on Build GRCh38
rs1022584295 Chr.11: 117824718-117824718 on Build GRCh38
rs12273102 Chr.11: 117824757-117824757 on Build GRCh38
rs563129744 Chr.11: 117824766-117824766 on Build GRCh38
rs983942834 Chr.11: 117824584-117824585 on Build GRCh38
rs755640340 Chr.11: 117824728-117824728 on Build GRCh38
rs375591023 Chr.11: 117824712-117824712 on Build GRCh38
rs749565936 Chr.11: 117824665-117824665 on Build GRCh38
rs368731453 Chr.11: 117824580-117824580 on Build GRCh38
rs113299959 Chr.11: 117824681-117824681 on Build GRCh38
rs753343885 Chr.11: 117824686-117824686 on Build GRCh38
rs3824943 Chr.11: 117824625-117824625 on Build GRCh38
rs768418229 Chr.11: 117824667-117824667 on Build GRCh38
rs191708738 Chr.11: 117824657-117824657 on Build GRCh38
rs781108382 Chr.11: 117824726-117824726 on Build GRCh38
rs369874877 Chr.11: 117824630-117824630 on Build GRCh38
rs12279985 Chr.11: 117824688-117824688 on Build GRCh38
rs966962819 Chr.11: 117824735-117824735 on Build GRCh38
rs760843021 Chr.11: 117824691-117824691 on Build GRCh38
rs747003225 Chr.11: 117824636-117824636 on Build GRCh38
rs767744240 Chr.11: 117824627-117824627 on Build GRCh38
rs747791835 Chr.11: 117824647-117824647 on Build GRCh38
rs745497369 Chr.11: 117824626-117824626 on Build GRCh38
rs766240606 Chr.11: 117824701-117824701 on Build GRCh38

View Full Product Details