Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93452313-93452556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120A
Entrez Gene ID
23196
Gene Name
family with sequence similarity 120A
Gene Aliases
C9orf10, OSSA
UniGene
Hs.372003
Gene Chromosome Location
Chr.9: 93451696-93566115 on Build GRCh38
Transcripts
NM_001286722.1 NM_001286723.1 NM_001286724.1 NM_014612.4 XM_005251842.4 XM_011518412.2 XM_011518413.2 XM_011518414.2 XM_011518416.2 NM_198841.2 NR_136229.1 NR_136230.1 NR_136231.1

Gene Symbol
FAM120AOS
Entrez Gene ID
158293
Gene Name
family with sequence similarity 120A opposite strand
Gene Aliases
C9orf10OS
UniGene
Hs.350364
Gene Chromosome Location
Chr.9: 93446500-93453592 on Build GRCh38
Transcripts
NM_001286722.1 NM_001286723.1 NM_001286724.1 NM_014612.4 XM_005251842.4 XM_011518412.2 XM_011518413.2 XM_011518414.2 XM_011518416.2 NM_198841.2 NR_136229.1 NR_136230.1 NR_136231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6116598 c.311C>A p.P104H Chr.9: 93452399-93452399 on Build GRCh38
1206045 c.394G>T p.G132C Chr.9: 93452316-93452316 on Build GRCh38
6116599 c.474+10G>T p.? Chr.9: 93452399-93452399 on Build GRCh38
5757946 c.320C>T p.T107I Chr.9: 93452390-93452390 on Build GRCh38
6116597 c.474+10G>T p.? Chr.9: 93452399-93452399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777538417 Chr.9: 93452394-93452410 on Build GRCh38
rs756034568 Chr.9: 93452476-93452476 on Build GRCh38
rs372272292 Chr.9: 93452397-93452397 on Build GRCh38
rs564664349 Chr.9: 93452526-93452526 on Build GRCh38
rs765234116 Chr.9: 93452454-93452454 on Build GRCh38
rs865982421 Chr.9: 93452351-93452351 on Build GRCh38
rs748898779 Chr.9: 93452491-93452491 on Build GRCh38
rs1033250405 Chr.9: 93452409-93452410 on Build GRCh38
rs767466439 Chr.9: 93452504-93452504 on Build GRCh38
rs925216346 Chr.9: 93452513-93452513 on Build GRCh38
rs746376434 Chr.9: 93452425-93452425 on Build GRCh38
rs767607276 Chr.9: 93452469-93452469 on Build GRCh38
rs1013288049 Chr.9: 93452427-93452427 on Build GRCh38
rs745343672 Chr.9: 93452516-93452516 on Build GRCh38
rs757329043 Chr.9: 93452338-93452338 on Build GRCh38
rs754331377 Chr.9: 93452468-93452468 on Build GRCh38
rs752664497 Chr.9: 93452470-93452470 on Build GRCh38
rs772520926 Chr.9: 93452430-93452430 on Build GRCh38
rs754874835 Chr.9: 93452354-93452354 on Build GRCh38
rs937037051 Chr.9: 93452507-93452507 on Build GRCh38
rs770475308 Chr.9: 93452424-93452425 on Build GRCh38
rs776648143 Chr.9: 93452440-93452440 on Build GRCh38
rs773846801 Chr.9: 93452422-93452422 on Build GRCh38
rs764419052 Chr.9: 93452439-93452439 on Build GRCh38
rs777708984 Chr.9: 93452488-93452488 on Build GRCh38
rs868811104 Chr.9: 93452348-93452348 on Build GRCh38
rs372504822 Chr.9: 93452517-93452517 on Build GRCh38
rs747899074 Chr.9: 93452383-93452383 on Build GRCh38
rs200709426 Chr.9: 93452531-93452531 on Build GRCh38
rs751515563 Chr.9: 93452349-93452349 on Build GRCh38
rs765344986 Chr.9: 93452345-93452345 on Build GRCh38
rs770417375 Chr.9: 93452415-93452415 on Build GRCh38
rs762088381 Chr.9: 93452445-93452445 on Build GRCh38
rs1023278688 Chr.9: 93452473-93452473 on Build GRCh38
rs755946786 Chr.9: 93452384-93452384 on Build GRCh38
rs1037874200 Chr.9: 93452335-93452335 on Build GRCh38
rs971938855 Chr.9: 93452496-93452496 on Build GRCh38
rs756879211 Chr.9: 93452499-93452499 on Build GRCh38
rs776773791 Chr.9: 93452443-93452443 on Build GRCh38
rs368893571 Chr.9: 93452374-93452374 on Build GRCh38
rs769075234 Chr.9: 93452539-93452539 on Build GRCh38
rs544670498 Chr.9: 93452438-93452438 on Build GRCh38
rs988977140 Chr.9: 93452448-93452448 on Build GRCh38
rs771538981 Chr.9: 93452520-93452520 on Build GRCh38
rs927012067 Chr.9: 93452501-93452501 on Build GRCh38
rs913392014 Chr.9: 93452450-93452450 on Build GRCh38
rs748960781 Chr.9: 93452406-93452406 on Build GRCh38
rs372565267 Chr.9: 93452467-93452467 on Build GRCh38
rs746494654 Chr.9: 93452404-93452420 on Build GRCh38
rs553868255 Chr.9: 93452508-93452508 on Build GRCh38
rs750455274 Chr.9: 93452457-93452457 on Build GRCh38

View Full Product Details