Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74343564-74343837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11A1
Entrez Gene ID
1583
Gene Name
cytochrome P450 family 11 subfamily A member 1
Gene Aliases
CYP11A, CYPXIA1, P450SCC
UniGene
Hs.303980
Gene Chromosome Location
Chr.15: 74337762-74367740 on Build GRCh38
Transcripts
NM_000781.2 NM_001099773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1517490 c.814G>T p.V272L Chr.15: 74343804-74343804 on Build GRCh38
7267510 c.829+8C>T p.? Chr.15: 74343781-74343781 on Build GRCh38
81065 c.798T>C p.H266H Chr.15: 74343820-74343820 on Build GRCh38
1938466 c.829+1G>T p.? Chr.15: 74343788-74343788 on Build GRCh38
4056866 c.814G>A p.V272M Chr.15: 74343804-74343804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527958779 Chr.15: 74343738-74343738 on Build GRCh38
rs766748410 Chr.15: 74343673-74343673 on Build GRCh38
rs1053782077 Chr.15: 74343659-74343659 on Build GRCh38
rs377347885 Chr.15: 74343765-74343765 on Build GRCh38
rs183718296 Chr.15: 74343757-74343757 on Build GRCh38
rs921345473 Chr.15: 74343675-74343675 on Build GRCh38
rs750547647 Chr.15: 74343804-74343804 on Build GRCh38
rs113626801 Chr.15: 74343726-74343726 on Build GRCh38
rs560266518 Chr.15: 74343706-74343706 on Build GRCh38
rs934111926 Chr.15: 74343717-74343717 on Build GRCh38
rs1037625136 Chr.15: 74343609-74343609 on Build GRCh38
rs192766301 Chr.15: 74343661-74343661 on Build GRCh38
rs936181378 Chr.15: 74343667-74343667 on Build GRCh38
rs978096968 Chr.15: 74343610-74343610 on Build GRCh38
rs376847692 Chr.15: 74343772-74343772 on Build GRCh38
rs898286504 Chr.15: 74343756-74343756 on Build GRCh38
rs1130843 Chr.15: 74343796-74343796 on Build GRCh38
rs369873220 Chr.15: 74343786-74343786 on Build GRCh38
rs751908046 Chr.15: 74343722-74343722 on Build GRCh38
rs926116271 Chr.15: 74343627-74343627 on Build GRCh38
rs751460047 Chr.15: 74343760-74343760 on Build GRCh38
rs560754203 Chr.15: 74343733-74343733 on Build GRCh38
rs1038700781 Chr.15: 74343737-74343737 on Build GRCh38
rs371896933 Chr.15: 74343805-74343805 on Build GRCh38
rs1021942880 Chr.15: 74343812-74343812 on Build GRCh38
rs200847366 Chr.15: 74343785-74343785 on Build GRCh38
rs556521060 Chr.15: 74343595-74343595 on Build GRCh38
rs574806239 Chr.15: 74343721-74343721 on Build GRCh38

View Full Product Details