Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 169660283-169660552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN3
Entrez Gene ID
1182
Gene Name
chloride voltage-gated channel 3
Gene Aliases
CLC3, ClC-3
UniGene
Hs.481186
Gene Chromosome Location
Chr.4: 169620521-169723187 on Build GRCh38
Transcripts
NM_001243372.1 NM_001243374.1 NM_001829.3 NM_173872.3 XM_005262726.2

SNP Detail

SNP ID Chromosome Location
rs1019186622 Chr.4: 169660364-169660364 on Build GRCh38
rs557887565 Chr.4: 169660505-169660505 on Build GRCh38
rs1054912376 Chr.4: 169660524-169660524 on Build GRCh38
rs142659889 Chr.4: 169660513-169660513 on Build GRCh38
rs780203659 Chr.4: 169660349-169660349 on Build GRCh38
rs1029199914 Chr.4: 169660381-169660381 on Build GRCh38
rs902123435 Chr.4: 169660371-169660371 on Build GRCh38
rs891750060 Chr.4: 169660322-169660322 on Build GRCh38
rs754487672 Chr.4: 169660344-169660344 on Build GRCh38
rs747270956 Chr.4: 169660354-169660354 on Build GRCh38
rs35527578 Chr.4: 169660374-169660375 on Build GRCh38
rs537683697 Chr.4: 169660499-169660499 on Build GRCh38
rs1035454971 Chr.4: 169660436-169660436 on Build GRCh38
rs1025279949 Chr.4: 169660368-169660368 on Build GRCh38
rs565298994 Chr.4: 169660376-169660376 on Build GRCh38
rs4692739 Chr.4: 169660419-169660419 on Build GRCh38
rs780786957 Chr.4: 169660413-169660413 on Build GRCh38
rs191793461 Chr.4: 169660435-169660435 on Build GRCh38
rs948000786 Chr.4: 169660496-169660496 on Build GRCh38
rs996335335 Chr.4: 169660380-169660380 on Build GRCh38
rs781669163 Chr.4: 169660421-169660421 on Build GRCh38
rs951826949 Chr.4: 169660384-169660384 on Build GRCh38
rs556703252 Chr.4: 169660476-169660476 on Build GRCh38
rs571999855 Chr.4: 169660497-169660497 on Build GRCh38
rs980562537 Chr.4: 169660502-169660502 on Build GRCh38
rs77021636 Chr.4: 169660441-169660441 on Build GRCh38
rs551534842 Chr.4: 169660310-169660318 on Build GRCh38
rs529749343 Chr.4: 169660414-169660414 on Build GRCh38
rs936518856 Chr.4: 169660514-169660514 on Build GRCh38

View Full Product Details