Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139541017-139541256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F9
Entrez Gene ID
2158
Gene Name
coagulation factor IX
Gene Aliases
F9 p22, FIX, HEMB, P19, PTC, THPH8
UniGene
Hs.522798
Gene Chromosome Location
Chr.X: 139530720-139563459 on Build GRCh38
Transcripts
NM_000133.3 NM_001313913.1 XM_005262397.4

SNP Detail

SNP ID Chromosome Location
rs137852274 Chr.X: 139541126-139541126 on Build GRCh38
rs398122990 Chr.X: 139541073-139541073 on Build GRCh38
rs6049 Chr.X: 139541196-139541196 on Build GRCh38
rs370571441 Chr.X: 139541044-139541044 on Build GRCh38
rs137852230 Chr.X: 139541076-139541076 on Build GRCh38
rs375734226 Chr.X: 139541199-139541199 on Build GRCh38
rs757434127 Chr.X: 139541113-139541113 on Build GRCh38
rs137852232 Chr.X: 139541099-139541099 on Build GRCh38
rs3134809 Chr.X: 139541049-139541049 on Build GRCh38
rs749754992 Chr.X: 139541193-139541193 on Build GRCh38
rs377592837 Chr.X: 139541118-139541118 on Build GRCh38
rs746148956 Chr.X: 139541138-139541138 on Build GRCh38
rs376942649 Chr.X: 139541043-139541043 on Build GRCh38
rs780435919 Chr.X: 139541182-139541182 on Build GRCh38
rs368156653 Chr.X: 139541225-139541225 on Build GRCh38
rs770120402 Chr.X: 139541162-139541162 on Build GRCh38
rs137852234 Chr.X: 139541127-139541127 on Build GRCh38
rs137852231 Chr.X: 139541085-139541085 on Build GRCh38
rs1054992012 Chr.X: 139541060-139541060 on Build GRCh38
rs751604122 Chr.X: 139541092-139541092 on Build GRCh38
rs137852233 Chr.X: 139541114-139541114 on Build GRCh38
rs755062115 Chr.X: 139541218-139541218 on Build GRCh38
rs1032331792 Chr.X: 139541053-139541053 on Build GRCh38
rs139289585 Chr.X: 139541131-139541131 on Build GRCh38

View Full Product Details