Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84372327-84372580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCSER2
Entrez Gene ID
54462
Gene Name
coiled-coil serine rich protein 2
Gene Aliases
FAM190B, Gcap14, KIAA1128, bA486O22.1
UniGene
Hs.461988
Gene Chromosome Location
Chr.10: 84328558-84518521 on Build GRCh38
Transcripts
NM_001284240.1 NM_001284241.1 NM_018999.3 XM_005269905.3 XM_005269906.3 XM_011539870.2 XM_011539871.2 XM_011539872.2 XM_017016340.1 XM_017016341.1 XM_017016342.1 XM_017016343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466022 c.1397G>A p.W466* Chr.10: 84372449-84372449 on Build GRCh38
3732176 c.1334C>A p.P445Q Chr.10: 84372386-84372386 on Build GRCh38
7418481 c.1417+2T>A p.? Chr.10: 84372471-84372471 on Build GRCh38
4393640 c.1376C>G p.A459G Chr.10: 84372428-84372428 on Build GRCh38
4682835 c.1374A>C p.K458N Chr.10: 84372426-84372426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769175420 Chr.10: 84372382-84372382 on Build GRCh38
rs988185421 Chr.10: 84372528-84372528 on Build GRCh38
rs759037265 Chr.10: 84372408-84372408 on Build GRCh38
rs772979371 Chr.10: 84372393-84372393 on Build GRCh38
rs777068231 Chr.10: 84372386-84372386 on Build GRCh38
rs199828450 Chr.10: 84372396-84372396 on Build GRCh38
rs977768306 Chr.10: 84372490-84372491 on Build GRCh38
rs747075612 Chr.10: 84372480-84372480 on Build GRCh38
rs762174831 Chr.10: 84372388-84372388 on Build GRCh38
rs140960423 Chr.10: 84372392-84372392 on Build GRCh38
rs756038809 Chr.10: 84372431-84372431 on Build GRCh38
rs1031928911 Chr.10: 84372526-84372527 on Build GRCh38
rs767574887 Chr.10: 84372424-84372424 on Build GRCh38
rs779279881 Chr.10: 84372365-84372365 on Build GRCh38
rs183950112 Chr.10: 84372447-84372447 on Build GRCh38
rs113299485 Chr.10: 84372462-84372462 on Build GRCh38
rs767771507 Chr.10: 84372504-84372504 on Build GRCh38
rs765949317 Chr.10: 84372399-84372399 on Build GRCh38
rs144817467 Chr.10: 84372407-84372407 on Build GRCh38
rs752613460 Chr.10: 84372428-84372428 on Build GRCh38
rs747072616 Chr.10: 84372377-84372377 on Build GRCh38
rs990780440 Chr.10: 84372548-84372548 on Build GRCh38
rs758580416 Chr.10: 84372461-84372461 on Build GRCh38
rs376616118 Chr.10: 84372373-84372373 on Build GRCh38
rs966427126 Chr.10: 84372487-84372487 on Build GRCh38
rs147804648 Chr.10: 84372458-84372458 on Build GRCh38
rs755915975 Chr.10: 84372357-84372357 on Build GRCh38
rs768525751 Chr.10: 84372374-84372374 on Build GRCh38
rs780603686 Chr.10: 84372482-84372483 on Build GRCh38
rs540535048 Chr.10: 84372529-84372529 on Build GRCh38
rs369899642 Chr.10: 84372484-84372484 on Build GRCh38
rs746071944 Chr.10: 84372371-84372371 on Build GRCh38

View Full Product Details