Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81526993-81527220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP12
Entrez Gene ID
646486
Gene Name
fatty acid binding protein 12
Gene Aliases
-
UniGene
Hs.586370
Gene Chromosome Location
Chr.8: 81524673-81590664 on Build GRCh38
Transcripts
NM_001105281.1 XM_005251285.4 XM_006716465.3 XM_011517577.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1569540 c.291C>A p.D97E Chr.8: 81527077-81527077 on Build GRCh38
4600428 c.313A>C p.I105L Chr.8: 81527055-81527055 on Build GRCh38
3651315 c.318G>A p.T106T Chr.8: 81527050-81527050 on Build GRCh38
3651313 c.324G>A p.K108K Chr.8: 81527044-81527044 on Build GRCh38
3835066 c.304G>A p.E102K Chr.8: 81527064-81527064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773076749 Chr.8: 81527022-81527022 on Build GRCh38
rs752261434 Chr.8: 81527075-81527075 on Build GRCh38
rs371573132 Chr.8: 81527054-81527054 on Build GRCh38
rs568288088 Chr.8: 81527051-81527051 on Build GRCh38
rs770211701 Chr.8: 81527014-81527014 on Build GRCh38
rs185223281 Chr.8: 81527105-81527105 on Build GRCh38
rs956359181 Chr.8: 81527033-81527033 on Build GRCh38
rs766991525 Chr.8: 81527069-81527069 on Build GRCh38
rs868782483 Chr.8: 81527037-81527037 on Build GRCh38
rs766179971 Chr.8: 81527032-81527032 on Build GRCh38
rs773505573 Chr.8: 81527170-81527170 on Build GRCh38
rs368067152 Chr.8: 81527050-81527050 on Build GRCh38
rs980285877 Chr.8: 81527084-81527084 on Build GRCh38
rs574975609 Chr.8: 81527088-81527088 on Build GRCh38
rs758809104 Chr.8: 81527096-81527096 on Build GRCh38
rs866183763 Chr.8: 81527123-81527123 on Build GRCh38
rs752950072 Chr.8: 81527073-81527073 on Build GRCh38
rs371779914 Chr.8: 81527154-81527154 on Build GRCh38
rs375883973 Chr.8: 81527085-81527085 on Build GRCh38
rs941463615 Chr.8: 81527089-81527089 on Build GRCh38
rs535737231 Chr.8: 81527120-81527120 on Build GRCh38
rs781066160 Chr.8: 81527171-81527171 on Build GRCh38
rs564575673 Chr.8: 81527086-81527086 on Build GRCh38
rs781419689 Chr.8: 81527111-81527111 on Build GRCh38
rs779067892 Chr.8: 81527070-81527070 on Build GRCh38
rs747532400 Chr.8: 81527104-81527104 on Build GRCh38
rs770085173 Chr.8: 81527116-81527116 on Build GRCh38
rs748244972 Chr.8: 81527115-81527115 on Build GRCh38
rs780381787 Chr.8: 81527098-81527098 on Build GRCh38
rs375525557 Chr.8: 81527025-81527025 on Build GRCh38
rs763400246 Chr.8: 81527111-81527112 on Build GRCh38

View Full Product Details