Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63024097-63024338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGH
Entrez Gene ID
8836
Gene Name
gamma-glutamyl hydrolase
Gene Aliases
GH
UniGene
Hs.78619
Gene Chromosome Location
Chr.8: 63015079-63039051 on Build GRCh38
Transcripts
NM_003878.2 XM_011517623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7369412 c.584A>G p.H195R Chr.8: 63024102-63024102 on Build GRCh38
5630200 c.538G>C p.E180Q Chr.8: 63024148-63024148 on Build GRCh38
7081919 c.525G>C p.Q175H Chr.8: 63024161-63024161 on Build GRCh38
4686858 c.577A>G p.N193D Chr.8: 63024109-63024109 on Build GRCh38
3951766 c.507G>T p.L169F Chr.8: 63024179-63024179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749835115 Chr.8: 63024207-63024207 on Build GRCh38
rs73684329 Chr.8: 63024305-63024305 on Build GRCh38
rs1038862303 Chr.8: 63024307-63024307 on Build GRCh38
rs1039840031 Chr.8: 63024265-63024265 on Build GRCh38
rs755199444 Chr.8: 63024164-63024164 on Build GRCh38
rs371749964 Chr.8: 63024175-63024175 on Build GRCh38
rs986960195 Chr.8: 63024151-63024151 on Build GRCh38
rs140077692 Chr.8: 63024126-63024126 on Build GRCh38
rs778705774 Chr.8: 63024161-63024161 on Build GRCh38
rs369683727 Chr.8: 63024138-63024138 on Build GRCh38
rs771364585 Chr.8: 63024220-63024220 on Build GRCh38
rs753850058 Chr.8: 63024140-63024140 on Build GRCh38
rs935609323 Chr.8: 63024149-63024149 on Build GRCh38
rs989496297 Chr.8: 63024127-63024127 on Build GRCh38
rs370633636 Chr.8: 63024281-63024281 on Build GRCh38
rs911315574 Chr.8: 63024293-63024293 on Build GRCh38
rs747178839 Chr.8: 63024163-63024163 on Build GRCh38
rs773562149 Chr.8: 63024176-63024176 on Build GRCh38
rs924150363 Chr.8: 63024139-63024139 on Build GRCh38
rs898260100 Chr.8: 63024277-63024277 on Build GRCh38
rs781545152 Chr.8: 63024167-63024167 on Build GRCh38
rs1051139112 Chr.8: 63024269-63024269 on Build GRCh38
rs748165012 Chr.8: 63024169-63024169 on Build GRCh38
rs757009855 Chr.8: 63024156-63024156 on Build GRCh38
rs755704305 Chr.8: 63024129-63024129 on Build GRCh38
rs761998785 Chr.8: 63024224-63024225 on Build GRCh38

View Full Product Details