Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56397284-56397527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF1
Entrez Gene ID
1477
Gene Name
cleavage stimulation factor subunit 1
Gene Aliases
CstF-50, CstFp50
UniGene
Hs.172865
Gene Chromosome Location
Chr.20: 56392371-56404526 on Build GRCh38
Transcripts
NM_001033521.1 NM_001033522.1 NM_001324.2 XM_011528600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637022 c.298G>A p.E100K Chr.20: 56397335-56397335 on Build GRCh38
1734883 c.396A>G p.I132M Chr.20: 56397433-56397433 on Build GRCh38
4618047 c.420G>A p.M140I Chr.20: 56397457-56397457 on Build GRCh38
7279147 c.320_322delATA p.H107_K108>Q Chr.20: 56397357-56397359 on Build GRCh38
6637026 c.377G>A p.G126E Chr.20: 56397414-56397414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772433993 Chr.20: 56397451-56397451 on Build GRCh38
rs6024849 Chr.20: 56397364-56397364 on Build GRCh38
rs760868937 Chr.20: 56397479-56397479 on Build GRCh38
rs768748704 Chr.20: 56397491-56397491 on Build GRCh38
rs777217695 Chr.20: 56397404-56397404 on Build GRCh38
rs775607803 Chr.20: 56397331-56397331 on Build GRCh38
rs185193577 Chr.20: 56397430-56397430 on Build GRCh38
rs780923152 Chr.20: 56397380-56397380 on Build GRCh38
rs111908795 Chr.20: 56397403-56397403 on Build GRCh38
rs779423104 Chr.20: 56397427-56397427 on Build GRCh38
rs749772745 Chr.20: 56397406-56397406 on Build GRCh38
rs764134713 Chr.20: 56397349-56397349 on Build GRCh38
rs567844389 Chr.20: 56397315-56397315 on Build GRCh38
rs138767968 Chr.20: 56397458-56397458 on Build GRCh38
rs767682335 Chr.20: 56397329-56397329 on Build GRCh38
rs142768610 Chr.20: 56397334-56397334 on Build GRCh38
rs766250406 Chr.20: 56397367-56397367 on Build GRCh38
rs1019213052 Chr.20: 56397319-56397319 on Build GRCh38
rs551234706 Chr.20: 56397311-56397311 on Build GRCh38
rs372533821 Chr.20: 56397496-56397496 on Build GRCh38
rs751350281 Chr.20: 56397372-56397372 on Build GRCh38
rs750367433 Chr.20: 56397352-56397352 on Build GRCh38
rs752305246 Chr.20: 56397400-56397400 on Build GRCh38
rs771477732 Chr.20: 56397420-56397420 on Build GRCh38
rs759735953 Chr.20: 56397320-56397320 on Build GRCh38
rs754851634 Chr.20: 56397379-56397379 on Build GRCh38

View Full Product Details