Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 443221-443468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM101B
Entrez Gene ID
359845
Gene Name
family with sequence similarity 101 member B
Gene Aliases
CFM1
UniGene
Hs.345588
Gene Chromosome Location
Chr.17: 439978-445940 on Build GRCh38
Transcripts
NM_182705.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
705689 c.268C>G p.P90A Chr.17: 443331-443331 on Build GRCh38
3179515 c.259C>T p.R87C Chr.17: 443340-443340 on Build GRCh38
3179519 c.227G>A p.R76H Chr.17: 443372-443372 on Build GRCh38
7096857 c.144C>A p.F48L Chr.17: 443455-443455 on Build GRCh38
3179520 c.222G>A p.T74T Chr.17: 443377-443377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781865189 Chr.17: 443433-443433 on Build GRCh38
rs782771305 Chr.17: 443430-443430 on Build GRCh38
rs370609692 Chr.17: 443358-443358 on Build GRCh38
rs781929540 Chr.17: 443270-443270 on Build GRCh38
rs782078204 Chr.17: 443271-443271 on Build GRCh38
rs562456865 Chr.17: 443258-443258 on Build GRCh38
rs963800067 Chr.17: 443391-443391 on Build GRCh38
rs201196510 Chr.17: 443280-443280 on Build GRCh38
rs781798139 Chr.17: 443293-443293 on Build GRCh38
rs781985104 Chr.17: 443402-443402 on Build GRCh38
rs374226782 Chr.17: 443340-443340 on Build GRCh38
rs782216464 Chr.17: 443393-443393 on Build GRCh38
rs373345868 Chr.17: 443284-443284 on Build GRCh38
rs782148415 Chr.17: 443281-443281 on Build GRCh38
rs74588575 Chr.17: 443255-443255 on Build GRCh38
rs782031541 Chr.17: 443419-443419 on Build GRCh38
rs782097171 Chr.17: 443345-443345 on Build GRCh38
rs781845685 Chr.17: 443304-443304 on Build GRCh38
rs782767576 Chr.17: 443346-443346 on Build GRCh38
rs368788591 Chr.17: 443401-443401 on Build GRCh38
rs782131752 Chr.17: 443404-443404 on Build GRCh38
rs571620470 Chr.17: 443329-443329 on Build GRCh38
rs8069166 Chr.17: 443347-443347 on Build GRCh38
rs912160216 Chr.17: 443428-443428 on Build GRCh38
rs782792365 Chr.17: 443413-443413 on Build GRCh38
rs200262570 Chr.17: 443315-443315 on Build GRCh38
rs375022749 Chr.17: 443249-443249 on Build GRCh38
rs370299014 Chr.17: 443324-443324 on Build GRCh38
rs781967796 Chr.17: 443325-443325 on Build GRCh38
rs200382363 Chr.17: 443242-443242 on Build GRCh38
rs782360011 Chr.17: 443261-443262 on Build GRCh38
rs782220939 Chr.17: 443368-443368 on Build GRCh38
rs782057242 Chr.17: 443338-443338 on Build GRCh38
rs373394872 Chr.17: 443246-443246 on Build GRCh38
rs782050755 Chr.17: 443421-443421 on Build GRCh38
rs782472948 Chr.17: 443423-443423 on Build GRCh38
rs782465699 Chr.17: 443360-443360 on Build GRCh38
rs782713598 Chr.17: 443339-443339 on Build GRCh38
rs368165658 Chr.17: 443367-443367 on Build GRCh38
rs782508006 Chr.17: 443305-443305 on Build GRCh38
rs117842563 Chr.17: 443384-443384 on Build GRCh38
rs550861769 Chr.17: 443372-443372 on Build GRCh38
rs782276843 Chr.17: 443313-443313 on Build GRCh38
rs369163506 Chr.17: 443250-443250 on Build GRCh38
rs189011908 Chr.17: 443303-443303 on Build GRCh38
rs372868305 Chr.17: 443377-443377 on Build GRCh38
rs532873330 Chr.17: 443309-443309 on Build GRCh38
rs782740032 Chr.17: 443275-443275 on Build GRCh38
rs921978372 Chr.17: 443407-443407 on Build GRCh38
rs782159607 Chr.17: 443257-443257 on Build GRCh38
rs782557368 Chr.17: 443445-443445 on Build GRCh38
rs782575478 Chr.17: 443446-443446 on Build GRCh38
rs201027561 Chr.17: 443359-443359 on Build GRCh38
rs782532789 Chr.17: 443351-443351 on Build GRCh38
rs782411833 Chr.17: 443326-443326 on Build GRCh38
rs536237828 Chr.17: 443378-443378 on Build GRCh38
rs782206933 Chr.17: 443448-443448 on Build GRCh38
rs894394971 Chr.17: 443352-443352 on Build GRCh38
rs782646185 Chr.17: 443373-443373 on Build GRCh38

View Full Product Details