Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38961834-38962095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO47
Entrez Gene ID
494188
Gene Name
F-box protein 47
Gene Aliases
-
UniGene
Hs.549536
Gene Chromosome Location
Chr.17: 38936278-38967475 on Build GRCh38
Transcripts
NM_001008777.2 XM_011524865.2 XM_011524866.2 XM_011524867.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
978518 c.341A>G p.Y114C Chr.17: 38961888-38961888 on Build GRCh38
5386195 c.194G>A p.S65N Chr.17: 38962035-38962035 on Build GRCh38
978519 c.293A>G p.N98S Chr.17: 38961936-38961936 on Build GRCh38
4579632 c.212C>T p.S71F Chr.17: 38962017-38962017 on Build GRCh38
5386194 c.336G>A p.E112E Chr.17: 38961893-38961893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764053515 Chr.17: 38961963-38961963 on Build GRCh38
rs751302290 Chr.17: 38961867-38961867 on Build GRCh38
rs563432448 Chr.17: 38961953-38961953 on Build GRCh38
rs779526632 Chr.17: 38962064-38962064 on Build GRCh38
rs751510105 Chr.17: 38961905-38961906 on Build GRCh38
rs750141493 Chr.17: 38962000-38962000 on Build GRCh38
rs761097409 Chr.17: 38961965-38961965 on Build GRCh38
rs200266705 Chr.17: 38961939-38961939 on Build GRCh38
rs144695633 Chr.17: 38961987-38961987 on Build GRCh38
rs754602414 Chr.17: 38962060-38962061 on Build GRCh38
rs747603172 Chr.17: 38962066-38962067 on Build GRCh38
rs369741439 Chr.17: 38961999-38961999 on Build GRCh38
rs755658854 Chr.17: 38961885-38961885 on Build GRCh38
rs756890923 Chr.17: 38961868-38961868 on Build GRCh38
rs751333422 Chr.17: 38961969-38961969 on Build GRCh38
rs769145743 Chr.17: 38961900-38961900 on Build GRCh38
rs202147522 Chr.17: 38961913-38961913 on Build GRCh38
rs753319659 Chr.17: 38962048-38962048 on Build GRCh38
rs748501202 Chr.17: 38961909-38961909 on Build GRCh38
rs755467692 Chr.17: 38962061-38962061 on Build GRCh38
rs779533632 Chr.17: 38961893-38961893 on Build GRCh38
rs1031309799 Chr.17: 38961973-38961973 on Build GRCh38
rs780773341 Chr.17: 38961874-38961874 on Build GRCh38
rs199842302 Chr.17: 38961895-38961895 on Build GRCh38
rs779550209 Chr.17: 38962011-38962011 on Build GRCh38
rs761645079 Chr.17: 38961994-38961994 on Build GRCh38
rs771195291 Chr.17: 38961929-38961929 on Build GRCh38
rs755748378 Chr.17: 38962005-38962005 on Build GRCh38
rs772546631 Chr.17: 38961910-38961910 on Build GRCh38
rs774579041 Chr.17: 38961904-38961904 on Build GRCh38
rs745497407 Chr.17: 38961876-38961876 on Build GRCh38
rs1023222255 Chr.17: 38961903-38961903 on Build GRCh38
rs748577537 Chr.17: 38962065-38962065 on Build GRCh38
rs556669338 Chr.17: 38962063-38962063 on Build GRCh38

View Full Product Details