Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51950592-51950861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG2
Entrez Gene ID
54331
Gene Name
G protein subunit gamma 2
Gene Aliases
-
UniGene
Hs.187772
Gene Chromosome Location
Chr.14: 51847131-51969800 on Build GRCh38
Transcripts
NM_001243773.1 NM_001243774.1 NM_053064.4 XM_006720173.2 XM_011536846.2 XM_017021376.1 XM_017021377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419870 c.75C>T p.I25I Chr.14: 51950753-51950753 on Build GRCh38
6140422 c.81G>C p.R27S Chr.14: 51950759-51950759 on Build GRCh38
6140423 c.78C>A p.D26E Chr.14: 51950756-51950756 on Build GRCh38
5664066 c.42G>A p.K14K Chr.14: 51950720-51950720 on Build GRCh38
5984020 c.38G>A p.R13K Chr.14: 51950716-51950716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528265437 Chr.14: 51950817-51950817 on Build GRCh38
rs774310897 Chr.14: 51950742-51950742 on Build GRCh38
rs777396127 Chr.14: 51950678-51950678 on Build GRCh38
rs577143990 Chr.14: 51950794-51950794 on Build GRCh38
rs377488071 Chr.14: 51950804-51950804 on Build GRCh38
rs771997787 Chr.14: 51950754-51950754 on Build GRCh38
rs781427591 Chr.14: 51950665-51950665 on Build GRCh38
rs981342625 Chr.14: 51950648-51950648 on Build GRCh38
rs760453739 Chr.14: 51950760-51950760 on Build GRCh38
rs531961329 Chr.14: 51950647-51950647 on Build GRCh38
rs373428226 Chr.14: 51950799-51950799 on Build GRCh38
rs372811211 Chr.14: 51950811-51950811 on Build GRCh38
rs45501792 Chr.14: 51950627-51950627 on Build GRCh38
rs777449701 Chr.14: 51950816-51950816 on Build GRCh38
rs765442953 Chr.14: 51950631-51950631 on Build GRCh38
rs770919668 Chr.14: 51950618-51950618 on Build GRCh38
rs756485867 Chr.14: 51950814-51950814 on Build GRCh38
rs769332235 Chr.14: 51950674-51950674 on Build GRCh38
rs780767653 Chr.14: 51950637-51950637 on Build GRCh38
rs140301757 Chr.14: 51950753-51950753 on Build GRCh38
rs758549985 Chr.14: 51950636-51950636 on Build GRCh38
rs755187459 Chr.14: 51950799-51950800 on Build GRCh38
rs1039938291 Chr.14: 51950833-51950833 on Build GRCh38
rs986264760 Chr.14: 51950815-51950815 on Build GRCh38
rs373581574 Chr.14: 51950812-51950812 on Build GRCh38
rs763177209 Chr.14: 51950784-51950784 on Build GRCh38
rs962091300 Chr.14: 51950798-51950798 on Build GRCh38
rs761174043 Chr.14: 51950619-51950619 on Build GRCh38
rs748853029 Chr.14: 51950696-51950696 on Build GRCh38
rs775163309 Chr.14: 51950756-51950756 on Build GRCh38
rs368850368 Chr.14: 51950724-51950724 on Build GRCh38
rs766539497 Chr.14: 51950785-51950785 on Build GRCh38
rs562485249 Chr.14: 51950697-51950697 on Build GRCh38
rs770404535 Chr.14: 51950708-51950708 on Build GRCh38
rs759423982 Chr.14: 51950623-51950623 on Build GRCh38
rs752094346 Chr.14: 51950641-51950641 on Build GRCh38
rs555437697 Chr.14: 51950625-51950625 on Build GRCh38
rs748484489 Chr.14: 51950673-51950673 on Build GRCh38
rs193225666 Chr.14: 51950809-51950809 on Build GRCh38
rs1024742790 Chr.14: 51950795-51950795 on Build GRCh38
rs376195482 Chr.14: 51950818-51950818 on Build GRCh38
rs926935924 Chr.14: 51950668-51950668 on Build GRCh38

View Full Product Details