Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55748401-55748674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF11
Entrez Gene ID
10220
Gene Name
growth differentiation factor 11
Gene Aliases
BMP-11, BMP11
UniGene
Hs.600883
Gene Chromosome Location
Chr.12: 55743278-55757194 on Build GRCh38
Transcripts
NM_005811.3 XM_006719194.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956494 c.447G>A p.T149T Chr.12: 55748587-55748587 on Build GRCh38
6670193 c.457G>A p.V153I Chr.12: 55748597-55748597 on Build GRCh38
5689721 c.464C>T p.T155I Chr.12: 55748604-55748604 on Build GRCh38
6581253 c.480C>T p.L160L Chr.12: 55748620-55748620 on Build GRCh38
6895360 c.466G>C p.D156H Chr.12: 55748606-55748606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965378504 Chr.12: 55748522-55748522 on Build GRCh38
rs757754558 Chr.12: 55748604-55748604 on Build GRCh38
rs138729368 Chr.12: 55748433-55748433 on Build GRCh38
rs1047943698 Chr.12: 55748507-55748507 on Build GRCh38
rs181823578 Chr.12: 55748532-55748532 on Build GRCh38
rs573169208 Chr.12: 55748549-55748549 on Build GRCh38
rs781608209 Chr.12: 55748612-55748612 on Build GRCh38
rs764956109 Chr.12: 55748569-55748569 on Build GRCh38
rs761302493 Chr.12: 55748544-55748547 on Build GRCh38
rs757036730 Chr.12: 55748482-55748482 on Build GRCh38
rs550172523 Chr.12: 55748445-55748445 on Build GRCh38
rs879714161 Chr.12: 55748560-55748560 on Build GRCh38
rs111520487 Chr.12: 55748438-55748438 on Build GRCh38
rs1037942672 Chr.12: 55748608-55748608 on Build GRCh38
rs780444518 Chr.12: 55748640-55748640 on Build GRCh38
rs749922085 Chr.12: 55748599-55748602 on Build GRCh38
rs745740256 Chr.12: 55748565-55748565 on Build GRCh38
rs778217141 Chr.12: 55748588-55748588 on Build GRCh38
rs770076921 Chr.12: 55748627-55748627 on Build GRCh38
rs369681542 Chr.12: 55748544-55748544 on Build GRCh38
rs374221911 Chr.12: 55748580-55748580 on Build GRCh38
rs539006048 Chr.12: 55748479-55748479 on Build GRCh38
rs746286407 Chr.12: 55748620-55748620 on Build GRCh38
rs753289736 Chr.12: 55748587-55748587 on Build GRCh38
rs571579427 Chr.12: 55748447-55748447 on Build GRCh38
rs750238991 Chr.12: 55748553-55748553 on Build GRCh38
rs746758754 Chr.12: 55748437-55748437 on Build GRCh38
rs569084969 Chr.12: 55748569-55748569 on Build GRCh38
rs747524031 Chr.12: 55748599-55748599 on Build GRCh38
rs974353773 Chr.12: 55748436-55748436 on Build GRCh38
rs764072303 Chr.12: 55748537-55748537 on Build GRCh38
rs34238045 Chr.12: 55748638-55748639 on Build GRCh38

View Full Product Details