Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57608992-57609265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX3
Entrez Gene ID
196403
Gene Name
deltex E3 ubiquitin ligase 3
Gene Aliases
RNF154, deltex3
UniGene
Hs.32374
Gene Chromosome Location
Chr.12: 57604327-57609804 on Build GRCh38
Transcripts
NM_001286245.1 NM_001286246.1 NM_178502.3 XM_005268697.1 XM_005268698.1 XM_005268700.1 XM_005268703.1 XM_011538022.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243180 c.970T>C p.F324L Chr.12: 57609078-57609078 on Build GRCh38
7170998 c.1026G>C p.L342L Chr.12: 57609125-57609125 on Build GRCh38
3955014 c.970T>G p.F324V Chr.12: 57609078-57609078 on Build GRCh38
7243179 c.979T>C p.F327L Chr.12: 57609078-57609078 on Build GRCh38
5067019 c.969G>T p.L323L Chr.12: 57609077-57609077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770335264 Chr.12: 57609030-57609030 on Build GRCh38
rs748571053 Chr.12: 57609130-57609130 on Build GRCh38
rs781461491 Chr.12: 57609196-57609196 on Build GRCh38
rs775023789 Chr.12: 57609157-57609157 on Build GRCh38
rs543446533 Chr.12: 57609153-57609153 on Build GRCh38
rs750461540 Chr.12: 57609040-57609040 on Build GRCh38
rs765028357 Chr.12: 57609168-57609168 on Build GRCh38
rs760229420 Chr.12: 57609160-57609160 on Build GRCh38
rs761762964 Chr.12: 57609163-57609163 on Build GRCh38
rs780137012 Chr.12: 57609116-57609116 on Build GRCh38
rs554617288 Chr.12: 57609054-57609054 on Build GRCh38
rs369658430 Chr.12: 57609077-57609077 on Build GRCh38
rs751711793 Chr.12: 57609188-57609188 on Build GRCh38
rs1031287009 Chr.12: 57609044-57609044 on Build GRCh38
rs759035747 Chr.12: 57609048-57609048 on Build GRCh38
rs776406575 Chr.12: 57609161-57609161 on Build GRCh38
rs760324992 Chr.12: 57609070-57609070 on Build GRCh38
rs906726682 Chr.12: 57609035-57609035 on Build GRCh38
rs934630710 Chr.12: 57609180-57609180 on Build GRCh38
rs781371652 Chr.12: 57609125-57609125 on Build GRCh38
rs756208640 Chr.12: 57609222-57609222 on Build GRCh38
rs747228849 Chr.12: 57609119-57609119 on Build GRCh38
rs576106515 Chr.12: 57609055-57609055 on Build GRCh38
rs376137016 Chr.12: 57609073-57609073 on Build GRCh38
rs758319322 Chr.12: 57609185-57609185 on Build GRCh38
rs56281065 Chr.12: 57609197-57609197 on Build GRCh38
rs750365437 Chr.12: 57609108-57609108 on Build GRCh38
rs879200168 Chr.12: 57609151-57609151 on Build GRCh38
rs750353500 Chr.12: 57609176-57609176 on Build GRCh38
rs373841536 Chr.12: 57609086-57609086 on Build GRCh38
rs916944647 Chr.12: 57609211-57609211 on Build GRCh38
rs755235895 Chr.12: 57609123-57609123 on Build GRCh38
rs755377232 Chr.12: 57609170-57609170 on Build GRCh38
rs770139096 Chr.12: 57609131-57609131 on Build GRCh38
rs371838961 Chr.12: 57609239-57609239 on Build GRCh38
rs558517499 Chr.12: 57609186-57609186 on Build GRCh38
rs755039624 Chr.12: 57609193-57609193 on Build GRCh38
rs765319327 Chr.12: 57609101-57609101 on Build GRCh38
rs372954011 Chr.12: 57609235-57609235 on Build GRCh38
rs773596800 Chr.12: 57609134-57609134 on Build GRCh38
rs372861555 Chr.12: 57609016-57609016 on Build GRCh38
rs1045497102 Chr.12: 57609015-57609015 on Build GRCh38
rs978568996 Chr.12: 57609106-57609106 on Build GRCh38
rs773756780 Chr.12: 57609041-57609041 on Build GRCh38
rs749744479 Chr.12: 57609145-57609145 on Build GRCh38

View Full Product Details