Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 93246857-93247082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA7
Entrez Gene ID
2045
Gene Name
EPH receptor A7
Gene Aliases
EHK-3, EHK3, EK11, HEK11
UniGene
Hs.73962
Gene Chromosome Location
Chr.6: 93240020-93419687 on Build GRCh38
Transcripts
NM_001288629.1 NM_004440.3 XM_005248669.2 XM_017010365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6972788 c.2590_2591GG>CT p.G864L Chr.6: 93246927-93246928 on Build GRCh38
3876199 c.2561G>A p.R854H Chr.6: 93246957-93246957 on Build GRCh38
5608518 c.2563T>C p.L855L Chr.6: 93246955-93246955 on Build GRCh38
6971691 c.2639G>A p.R880K Chr.6: 93246879-93246879 on Build GRCh38
1292380 c.2657T>G p.I886R Chr.6: 93246861-93246861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926719791 Chr.6: 93246974-93246974 on Build GRCh38
rs923098531 Chr.6: 93246885-93246885 on Build GRCh38
rs776802108 Chr.6: 93246957-93246957 on Build GRCh38
rs750587756 Chr.6: 93246990-93246990 on Build GRCh38
rs745373107 Chr.6: 93246897-93246897 on Build GRCh38
rs756995977 Chr.6: 93247015-93247015 on Build GRCh38
rs777630770 Chr.6: 93247006-93247006 on Build GRCh38
rs200816651 Chr.6: 93246942-93246942 on Build GRCh38
rs1038362394 Chr.6: 93246970-93246970 on Build GRCh38
rs1056681731 Chr.6: 93247042-93247042 on Build GRCh38
rs755970343 Chr.6: 93246888-93246888 on Build GRCh38
rs113927523 Chr.6: 93246938-93246938 on Build GRCh38
rs538121970 Chr.6: 93246981-93246981 on Build GRCh38
rs986033484 Chr.6: 93247031-93247031 on Build GRCh38
rs1009699233 Chr.6: 93247052-93247052 on Build GRCh38
rs55780501 Chr.6: 93246929-93246929 on Build GRCh38
rs745418263 Chr.6: 93247030-93247030 on Build GRCh38
rs762098670 Chr.6: 93246973-93246973 on Build GRCh38
rs749035576 Chr.6: 93247007-93247007 on Build GRCh38
rs778519646 Chr.6: 93247017-93247017 on Build GRCh38
rs777383467 Chr.6: 93246889-93246889 on Build GRCh38
rs766527264 Chr.6: 93246996-93246996 on Build GRCh38
rs755948630 Chr.6: 93246998-93246998 on Build GRCh38
rs772613034 Chr.6: 93246914-93246914 on Build GRCh38
rs758521423 Chr.6: 93246992-93246992 on Build GRCh38
rs752705560 Chr.6: 93246997-93246997 on Build GRCh38
rs184471651 Chr.6: 93246987-93246987 on Build GRCh38
rs377619656 Chr.6: 93246931-93246931 on Build GRCh38
rs450238 Chr.6: 93247051-93247051 on Build GRCh38
rs938075199 Chr.6: 93247041-93247041 on Build GRCh38
rs201874177 Chr.6: 93246982-93246982 on Build GRCh38
rs374483308 Chr.6: 93246893-93246893 on Build GRCh38
rs748985745 Chr.6: 93246892-93246892 on Build GRCh38

View Full Product Details