Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73292199-73292462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR4
Entrez Gene ID
1961
Gene Name
early growth response 4
Gene Aliases
NGFI-C, NGFIC, PAT133
UniGene
Hs.3052
Gene Chromosome Location
Chr.2: 73290929-73293701 on Build GRCh38
Transcripts
NM_001965.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652369 c.925G>C p.A309P Chr.2: 73292302-73292302 on Build GRCh38
4095613 c.589G>A p.A197T Chr.2: 73292329-73292329 on Build GRCh38
3799168 c.855C>T p.V285V Chr.2: 73292372-73292372 on Build GRCh38
1284695 c.801C>T p.C267C Chr.2: 73292426-73292426 on Build GRCh38
4095617 c.540C>T p.P180P Chr.2: 73292378-73292378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567025204 Chr.2: 73292232-73292232 on Build GRCh38
rs763600939 Chr.2: 73292251-73292251 on Build GRCh38
rs759199487 Chr.2: 73292231-73292231 on Build GRCh38
rs138743995 Chr.2: 73292238-73292238 on Build GRCh38
rs114378699 Chr.2: 73292306-73292306 on Build GRCh38
rs779921623 Chr.2: 73292366-73292366 on Build GRCh38
rs756336662 Chr.2: 73292273-73292273 on Build GRCh38
rs761255136 Chr.2: 73292404-73292404 on Build GRCh38
rs762330340 Chr.2: 73292263-73292263 on Build GRCh38
rs750784966 Chr.2: 73292342-73292342 on Build GRCh38
rs748851873 Chr.2: 73292370-73292370 on Build GRCh38
rs779144627 Chr.2: 73292299-73292299 on Build GRCh38
rs540230405 Chr.2: 73292381-73292381 on Build GRCh38
rs985927800 Chr.2: 73292307-73292307 on Build GRCh38
rs773828662 Chr.2: 73292387-73292387 on Build GRCh38
rs564426644 Chr.2: 73292374-73292374 on Build GRCh38
rs753945411 Chr.2: 73292353-73292353 on Build GRCh38
rs781115397 Chr.2: 73292302-73292302 on Build GRCh38
rs768061069 Chr.2: 73292338-73292338 on Build GRCh38
rs575492472 Chr.2: 73292311-73292311 on Build GRCh38
rs115948271 Chr.2: 73292360-73292360 on Build GRCh38
rs766768025 Chr.2: 73292348-73292348 on Build GRCh38
rs753944993 Chr.2: 73292277-73292277 on Build GRCh38
rs755269819 Chr.2: 73292358-73292358 on Build GRCh38
rs777081650 Chr.2: 73292412-73292412 on Build GRCh38
rs766599825 Chr.2: 73292410-73292410 on Build GRCh38
rs375223327 Chr.2: 73292436-73292436 on Build GRCh38
rs757251398 Chr.2: 73292362-73292362 on Build GRCh38
rs534204781 Chr.2: 73292236-73292236 on Build GRCh38
rs754944169 Chr.2: 73292291-73292291 on Build GRCh38
rs573006419 Chr.2: 73292378-73292378 on Build GRCh38
rs757920610 Chr.2: 73292395-73292395 on Build GRCh38
rs550247699 Chr.2: 73292445-73292445 on Build GRCh38
rs776586470 Chr.2: 73292227-73292227 on Build GRCh38
rs972746033 Chr.2: 73292401-73292401 on Build GRCh38
rs574121123 Chr.2: 73292275-73292275 on Build GRCh38
rs769419715 Chr.2: 73292365-73292365 on Build GRCh38
rs761369176 Chr.2: 73292398-73292398 on Build GRCh38
rs772561843 Chr.2: 73292399-73292399 on Build GRCh38
rs761010361 Chr.2: 73292347-73292347 on Build GRCh38
rs768337178 Chr.2: 73292384-73292384 on Build GRCh38
rs768106332 Chr.2: 73292317-73292317 on Build GRCh38
rs1005190122 Chr.2: 73292230-73292230 on Build GRCh38
rs752005782 Chr.2: 73292252-73292252 on Build GRCh38
rs372554686 Chr.2: 73292426-73292426 on Build GRCh38
rs934565954 Chr.2: 73292411-73292411 on Build GRCh38
rs767829076 Chr.2: 73292271-73292271 on Build GRCh38
rs113117246 Chr.2: 73292438-73292438 on Build GRCh38
rs750793256 Chr.2: 73292272-73292272 on Build GRCh38
rs561516958 Chr.2: 73292419-73292419 on Build GRCh38
rs773882185 Chr.2: 73292322-73292322 on Build GRCh38
rs976911286 Chr.2: 73292298-73292298 on Build GRCh38
rs779091705 Chr.2: 73292359-73292359 on Build GRCh38
rs745710598 Chr.2: 73292303-73292303 on Build GRCh38
rs781060656 Chr.2: 73292363-73292363 on Build GRCh38
rs774940854 Chr.2: 73292308-73292308 on Build GRCh38
rs773523022 Chr.2: 73292402-73292402 on Build GRCh38
rs144909297 Chr.2: 73292300-73292300 on Build GRCh38
rs759814942 Chr.2: 73292418-73292418 on Build GRCh38
rs762277076 Chr.2: 73292325-73292325 on Build GRCh38
rs968610071 Chr.2: 73292334-73292334 on Build GRCh38

View Full Product Details