Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230243263-230243488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT2
Entrez Gene ID
2590
Gene Name
polypeptide N-acetylgalactosaminyltransferase 2
Gene Aliases
GalNAc-T2
UniGene
Hs.743964
Gene Chromosome Location
Chr.1: 230057789-230282130 on Build GRCh38
Transcripts
NM_001291866.1 NM_004481.4 XM_017000963.1 XM_017000964.1 XM_017000965.1 XM_017000966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738507 c.696C>G p.H232Q Chr.1: 230243394-230243394 on Build GRCh38
269224 c.703G>A p.E235K Chr.1: 230243401-230243401 on Build GRCh38
6209172 c.608-10G>A p.? Chr.1: 230243296-230243296 on Build GRCh38
5822442 c.682G>C p.E228Q Chr.1: 230243380-230243380 on Build GRCh38
5845803 c.610C>T p.L204F Chr.1: 230243308-230243308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774828688 Chr.1: 230243326-230243327 on Build GRCh38
rs768020566 Chr.1: 230243468-230243478 on Build GRCh38
rs142046356 Chr.1: 230243346-230243346 on Build GRCh38
rs954628887 Chr.1: 230243449-230243449 on Build GRCh38
rs760497016 Chr.1: 230243438-230243438 on Build GRCh38
rs774693718 Chr.1: 230243296-230243296 on Build GRCh38
rs775654688 Chr.1: 230243389-230243389 on Build GRCh38
rs112101154 Chr.1: 230243334-230243334 on Build GRCh38
rs772502318 Chr.1: 230243387-230243387 on Build GRCh38
rs764812990 Chr.1: 230243315-230243315 on Build GRCh38
rs145628006 Chr.1: 230243332-230243332 on Build GRCh38
rs968307551 Chr.1: 230243429-230243429 on Build GRCh38
rs376870425 Chr.1: 230243327-230243327 on Build GRCh38
rs189302597 Chr.1: 230243310-230243310 on Build GRCh38
rs151298982 Chr.1: 230243379-230243379 on Build GRCh38
rs750666004 Chr.1: 230243461-230243461 on Build GRCh38
rs370170142 Chr.1: 230243394-230243394 on Build GRCh38
rs755938044 Chr.1: 230243352-230243352 on Build GRCh38
rs765833016 Chr.1: 230243412-230243412 on Build GRCh38
rs749525530 Chr.1: 230243282-230243282 on Build GRCh38
rs761032669 Chr.1: 230243314-230243314 on Build GRCh38
rs757320940 Chr.1: 230243335-230243335 on Build GRCh38
rs375900752 Chr.1: 230243441-230243441 on Build GRCh38
rs764168215 Chr.1: 230243440-230243440 on Build GRCh38
rs200971686 Chr.1: 230243437-230243437 on Build GRCh38
rs111823613 Chr.1: 230243462-230243462 on Build GRCh38
rs113017328 Chr.1: 230243457-230243457 on Build GRCh38
rs777096608 Chr.1: 230243400-230243400 on Build GRCh38
rs986552210 Chr.1: 230243456-230243456 on Build GRCh38
rs570311189 Chr.1: 230243424-230243424 on Build GRCh38
rs747399816 Chr.1: 230243391-230243391 on Build GRCh38
rs901114273 Chr.1: 230243342-230243342 on Build GRCh38
rs757381636 Chr.1: 230243371-230243371 on Build GRCh38
rs762432658 Chr.1: 230243320-230243320 on Build GRCh38
rs746029098 Chr.1: 230243382-230243382 on Build GRCh38
rs139591495 Chr.1: 230243326-230243326 on Build GRCh38
rs772285690 Chr.1: 230243299-230243299 on Build GRCh38
rs140249782 Chr.1: 230243328-230243328 on Build GRCh38
rs772780493 Chr.1: 230243319-230243319 on Build GRCh38
rs774014070 Chr.1: 230243418-230243418 on Build GRCh38
rs777847839 Chr.1: 230243355-230243355 on Build GRCh38
rs771338113 Chr.1: 230243295-230243295 on Build GRCh38
rs201212164 Chr.1: 230243423-230243423 on Build GRCh38
rs759943545 Chr.1: 230243453-230243454 on Build GRCh38
rs762487680 Chr.1: 230243405-230243405 on Build GRCh38
rs199981857 Chr.1: 230243447-230243447 on Build GRCh38

View Full Product Details