Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41953647-41953872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD4
Entrez Gene ID
30844
Gene Name
EH domain containing 4
Gene Aliases
PAST4
UniGene
Hs.143703
Gene Chromosome Location
Chr.15: 41899440-41972557 on Build GRCh38
Transcripts
NM_139265.3 XM_017022100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
81123 c.399C>T p.N133N Chr.15: 41953778-41953778 on Build GRCh38
6326162 c.336C>A p.T112T Chr.15: 41953841-41953841 on Build GRCh38
1372882 c.389G>A p.R130H Chr.15: 41953788-41953788 on Build GRCh38
2186758 c.413G>A p.R138Q Chr.15: 41953764-41953764 on Build GRCh38
6652451 c.384C>G p.L128L Chr.15: 41953793-41953793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946848835 Chr.15: 41953686-41953686 on Build GRCh38
rs1051315476 Chr.15: 41953775-41953775 on Build GRCh38
rs748481753 Chr.15: 41953778-41953778 on Build GRCh38
rs775721419 Chr.15: 41953823-41953823 on Build GRCh38
rs200999190 Chr.15: 41953734-41953734 on Build GRCh38
rs542506240 Chr.15: 41953844-41953844 on Build GRCh38
rs536144546 Chr.15: 41953694-41953694 on Build GRCh38
rs376970234 Chr.15: 41953816-41953816 on Build GRCh38
rs369664238 Chr.15: 41953774-41953774 on Build GRCh38
rs962764656 Chr.15: 41953833-41953833 on Build GRCh38
rs750225783 Chr.15: 41953843-41953843 on Build GRCh38
rs757030643 Chr.15: 41953731-41953731 on Build GRCh38
rs1017125979 Chr.15: 41953812-41953812 on Build GRCh38
rs746506103 Chr.15: 41953764-41953764 on Build GRCh38
rs932217428 Chr.15: 41953765-41953765 on Build GRCh38
rs753610114 Chr.15: 41953720-41953720 on Build GRCh38
rs770079590 Chr.15: 41953788-41953788 on Build GRCh38
rs566379150 Chr.15: 41953772-41953772 on Build GRCh38
rs772209765 Chr.15: 41953822-41953822 on Build GRCh38
rs779763740 Chr.15: 41953747-41953747 on Build GRCh38
rs372903734 Chr.15: 41953805-41953805 on Build GRCh38
rs774459679 Chr.15: 41953809-41953809 on Build GRCh38
rs890059376 Chr.15: 41953793-41953793 on Build GRCh38
rs778108744 Chr.15: 41953789-41953789 on Build GRCh38
rs765186481 Chr.15: 41953838-41953838 on Build GRCh38
rs376762413 Chr.15: 41953738-41953738 on Build GRCh38
rs150106003 Chr.15: 41953837-41953837 on Build GRCh38
rs772815933 Chr.15: 41953777-41953777 on Build GRCh38
rs749853517 Chr.15: 41953792-41953792 on Build GRCh38
rs763598850 Chr.15: 41953809-41953809 on Build GRCh38
rs549739179 Chr.15: 41953737-41953737 on Build GRCh38
rs868744705 Chr.15: 41953807-41953807 on Build GRCh38

View Full Product Details