Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36504560-36504799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXRED2
Entrez Gene ID
80020
Gene Name
FAD dependent oxidoreductase domain containing 2
Gene Aliases
ERFAD
UniGene
Hs.387601
Gene Chromosome Location
Chr.22: 36487186-36507101 on Build GRCh38
Transcripts
NM_001102371.1 NM_024955.5 XM_017028941.1 XM_017028942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4807835 c.597G>A p.E199E Chr.22: 36504697-36504697 on Build GRCh38
4506376 c.718C>T p.H240Y Chr.22: 36504576-36504576 on Build GRCh38
5092982 c.652A>G p.I218V Chr.22: 36504642-36504642 on Build GRCh38
1742633 c.625G>T p.E209* Chr.22: 36504669-36504669 on Build GRCh38
5789652 c.671C>T p.S224L Chr.22: 36504623-36504623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772133480 Chr.22: 36504709-36504709 on Build GRCh38
rs756611081 Chr.22: 36504766-36504766 on Build GRCh38
rs746254833 Chr.22: 36504601-36504601 on Build GRCh38
rs142547783 Chr.22: 36504622-36504622 on Build GRCh38
rs757702452 Chr.22: 36504666-36504666 on Build GRCh38
rs373581384 Chr.22: 36504640-36504640 on Build GRCh38
rs773262454 Chr.22: 36504711-36504711 on Build GRCh38
rs779079459 Chr.22: 36504691-36504691 on Build GRCh38
rs762516113 Chr.22: 36504753-36504753 on Build GRCh38
rs185586253 Chr.22: 36504648-36504648 on Build GRCh38
rs749482571 Chr.22: 36504690-36504690 on Build GRCh38
rs748245109 Chr.22: 36504696-36504696 on Build GRCh38
rs752134663 Chr.22: 36504747-36504747 on Build GRCh38
rs146630197 Chr.22: 36504731-36504731 on Build GRCh38
rs756559486 Chr.22: 36504684-36504684 on Build GRCh38
rs752224512 Chr.22: 36504662-36504662 on Build GRCh38
rs754269375 Chr.22: 36504769-36504769 on Build GRCh38
rs756280822 Chr.22: 36504594-36504594 on Build GRCh38
rs61730825 Chr.22: 36504685-36504685 on Build GRCh38
rs759137008 Chr.22: 36504728-36504728 on Build GRCh38
rs957451733 Chr.22: 36504636-36504636 on Build GRCh38
rs568610182 Chr.22: 36504613-36504613 on Build GRCh38
rs760588709 Chr.22: 36504645-36504645 on Build GRCh38
rs754449607 Chr.22: 36504658-36504658 on Build GRCh38
rs761793946 Chr.22: 36504633-36504633 on Build GRCh38
rs930809464 Chr.22: 36504687-36504687 on Build GRCh38
rs61730824 Chr.22: 36504727-36504727 on Build GRCh38
rs9610522 Chr.22: 36504765-36504765 on Build GRCh38
rs754328312 Chr.22: 36504736-36504736 on Build GRCh38
rs753580365 Chr.22: 36504650-36504650 on Build GRCh38
rs750748402 Chr.22: 36504679-36504679 on Build GRCh38
rs1045719925 Chr.22: 36504746-36504746 on Build GRCh38
rs779347247 Chr.22: 36504775-36504775 on Build GRCh38
rs760718 Chr.22: 36504759-36504759 on Build GRCh38
rs1007082166 Chr.22: 36504631-36504631 on Build GRCh38
rs540094773 Chr.22: 36504724-36504724 on Build GRCh38
rs757788754 Chr.22: 36504589-36504589 on Build GRCh38
rs755186215 Chr.22: 36504774-36504774 on Build GRCh38
rs764684253 Chr.22: 36504660-36504660 on Build GRCh38
rs770795939 Chr.22: 36504725-36504725 on Build GRCh38
rs144857776 Chr.22: 36504632-36504632 on Build GRCh38
rs981792611 Chr.22: 36504699-36504699 on Build GRCh38
rs781723894 Chr.22: 36504678-36504678 on Build GRCh38
rs775848102 Chr.22: 36504621-36504621 on Build GRCh38
rs1051760796 Chr.22: 36504773-36504773 on Build GRCh38
rs986474391 Chr.22: 36504585-36504585 on Build GRCh38

View Full Product Details