Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31844592-31844861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXS1
Entrez Gene ID
2307
Gene Name
forkhead box S1
Gene Aliases
FKHL18, FREAC10
UniGene
Hs.516971
Gene Chromosome Location
Chr.20: 31844300-31845617 on Build GRCh38
Transcripts
NM_004118.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3545095 c.870C>T p.P290P Chr.20: 31844673-31844673 on Build GRCh38
5028678 c.682G>C p.E228Q Chr.20: 31844861-31844861 on Build GRCh38
6666620 c.709G>A p.V237I Chr.20: 31844834-31844834 on Build GRCh38
3545096 c.820C>T p.P274S Chr.20: 31844723-31844723 on Build GRCh38
6666617 c.753G>A p.R251R Chr.20: 31844790-31844790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756030354 Chr.20: 31844831-31844831 on Build GRCh38
rs781337813 Chr.20: 31844762-31844762 on Build GRCh38
rs2296917 Chr.20: 31844669-31844669 on Build GRCh38
rs754326887 Chr.20: 31844713-31844713 on Build GRCh38
rs368204439 Chr.20: 31844660-31844660 on Build GRCh38
rs767390919 Chr.20: 31844827-31844827 on Build GRCh38
rs766879429 Chr.20: 31844708-31844708 on Build GRCh38
rs6089095 Chr.20: 31844665-31844665 on Build GRCh38
rs762296896 Chr.20: 31844612-31844612 on Build GRCh38
rs764476984 Chr.20: 31844815-31844815 on Build GRCh38
rs371636562 Chr.20: 31844717-31844717 on Build GRCh38
rs199689145 Chr.20: 31844811-31844811 on Build GRCh38
rs554902299 Chr.20: 31844639-31844639 on Build GRCh38
rs760976078 Chr.20: 31844705-31844705 on Build GRCh38
rs1006581850 Chr.20: 31844617-31844617 on Build GRCh38
rs781207851 Chr.20: 31844680-31844680 on Build GRCh38
rs771005189 Chr.20: 31844675-31844675 on Build GRCh38
rs747444343 Chr.20: 31844794-31844794 on Build GRCh38
rs548158235 Chr.20: 31844663-31844663 on Build GRCh38
rs200828167 Chr.20: 31844761-31844761 on Build GRCh38
rs199984812 Chr.20: 31844796-31844796 on Build GRCh38
rs779986698 Chr.20: 31844834-31844834 on Build GRCh38
rs768317733 Chr.20: 31844692-31844692 on Build GRCh38
rs562381219 Chr.20: 31844651-31844651 on Build GRCh38
rs779249477 Chr.20: 31844658-31844658 on Build GRCh38
rs779463483 Chr.20: 31844779-31844779 on Build GRCh38
rs145853748 Chr.20: 31844825-31844825 on Build GRCh38
rs763023094 Chr.20: 31844813-31844813 on Build GRCh38
rs751403711 Chr.20: 31844752-31844752 on Build GRCh38
rs768209810 Chr.20: 31844791-31844791 on Build GRCh38
rs111730906 Chr.20: 31844768-31844768 on Build GRCh38
rs750970202 Chr.20: 31844614-31844614 on Build GRCh38
rs148893203 Chr.20: 31844683-31844683 on Build GRCh38
rs748755819 Chr.20: 31844836-31844836 on Build GRCh38
rs752778362 Chr.20: 31844732-31844732 on Build GRCh38
rs748785422 Chr.20: 31844782-31844782 on Build GRCh38
rs143348822 Chr.20: 31844636-31844636 on Build GRCh38
rs758570616 Chr.20: 31844744-31844744 on Build GRCh38
rs778081779 Chr.20: 31844745-31844745 on Build GRCh38
rs975768941 Chr.20: 31844812-31844812 on Build GRCh38
rs143468590 Chr.20: 31844686-31844686 on Build GRCh38
rs35433949 Chr.20: 31844622-31844623 on Build GRCh38
rs777158180 Chr.20: 31844805-31844805 on Build GRCh38
rs139927827 Chr.20: 31844613-31844613 on Build GRCh38
rs765725671 Chr.20: 31844810-31844810 on Build GRCh38
rs530575086 Chr.20: 31844687-31844687 on Build GRCh38
rs773591743 Chr.20: 31844699-31844699 on Build GRCh38
rs773930926 Chr.20: 31844792-31844792 on Build GRCh38
rs753931627 Chr.20: 31844638-31844638 on Build GRCh38
rs146758105 Chr.20: 31844682-31844682 on Build GRCh38
rs376373877 Chr.20: 31844718-31844718 on Build GRCh38
rs751819004 Chr.20: 31844823-31844823 on Build GRCh38
rs761205373 Chr.20: 31844629-31844629 on Build GRCh38
rs765094531 Chr.20: 31844770-31844771 on Build GRCh38

View Full Product Details