Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4761073-4761322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXK1
Entrez Gene ID
221937
Gene Name
forkhead box K1
Gene Aliases
FOXK1L
UniGene
Hs.487393
Gene Chromosome Location
Chr.7: 4644691-4771443 on Build GRCh38
Transcripts
NM_001037165.1 XM_011515191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431129 c.1786G>A p.V596I Chr.7: 4761153-4761153 on Build GRCh38
6666353 c.1762G>A p.V588M Chr.7: 4761195-4761195 on Build GRCh38
3263953 c.1843G>A p.G615S Chr.7: 4761276-4761276 on Build GRCh38
3263952 c.1909G>A p.G637S Chr.7: 4761276-4761276 on Build GRCh38
7262344 c.1920C>T p.Y640Y Chr.7: 4761287-4761287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752161636 Chr.7: 4761294-4761295 on Build GRCh38
rs182098207 Chr.7: 4761104-4761104 on Build GRCh38
rs754424913 Chr.7: 4761239-4761239 on Build GRCh38
rs778485181 Chr.7: 4761192-4761192 on Build GRCh38
rs145281144 Chr.7: 4761159-4761159 on Build GRCh38
rs200934388 Chr.7: 4761254-4761254 on Build GRCh38
rs771784174 Chr.7: 4761195-4761195 on Build GRCh38
rs201318459 Chr.7: 4761131-4761131 on Build GRCh38
rs770509587 Chr.7: 4761277-4761277 on Build GRCh38
rs750885684 Chr.7: 4761166-4761166 on Build GRCh38
rs370145399 Chr.7: 4761253-4761253 on Build GRCh38
rs780430781 Chr.7: 4761116-4761116 on Build GRCh38
rs754227955 Chr.7: 4761282-4761282 on Build GRCh38
rs780896615 Chr.7: 4761107-4761107 on Build GRCh38
rs114866460 Chr.7: 4761298-4761298 on Build GRCh38
rs943691716 Chr.7: 4761175-4761175 on Build GRCh38
rs765180871 Chr.7: 4761205-4761206 on Build GRCh38
rs367809501 Chr.7: 4761231-4761231 on Build GRCh38
rs753426616 Chr.7: 4761288-4761288 on Build GRCh38
rs762904756 Chr.7: 4761155-4761155 on Build GRCh38
rs748714663 Chr.7: 4761185-4761185 on Build GRCh38
rs533152328 Chr.7: 4761203-4761203 on Build GRCh38
rs199885308 Chr.7: 4761280-4761280 on Build GRCh38
rs565840410 Chr.7: 4761186-4761186 on Build GRCh38
rs751285223 Chr.7: 4761106-4761106 on Build GRCh38
rs781694451 Chr.7: 4761164-4761164 on Build GRCh38
rs778395240 Chr.7: 4761243-4761243 on Build GRCh38
rs140406588 Chr.7: 4761095-4761095 on Build GRCh38
rs751855164 Chr.7: 4761158-4761158 on Build GRCh38
rs752595661 Chr.7: 4761150-4761150 on Build GRCh38
rs750176203 Chr.7: 4761276-4761276 on Build GRCh38
rs762894907 Chr.7: 4761204-4761204 on Build GRCh38
rs561545279 Chr.7: 4761128-4761128 on Build GRCh38
rs145875348 Chr.7: 4761264-4761264 on Build GRCh38
rs749746434 Chr.7: 4761118-4761118 on Build GRCh38
rs758137194 Chr.7: 4761101-4761101 on Build GRCh38
rs747808350 Chr.7: 4761193-4761193 on Build GRCh38
rs141919434 Chr.7: 4761230-4761230 on Build GRCh38
rs774659901 Chr.7: 4761127-4761127 on Build GRCh38
rs760570650 Chr.7: 4761201-4761201 on Build GRCh38
rs765076782 Chr.7: 4761097-4761097 on Build GRCh38
rs777445712 Chr.7: 4761262-4761262 on Build GRCh38
rs555700334 Chr.7: 4761275-4761275 on Build GRCh38
rs755469206 Chr.7: 4761287-4761287 on Build GRCh38
rs761395734 Chr.7: 4761242-4761242 on Build GRCh38
rs138556737 Chr.7: 4761281-4761281 on Build GRCh38
rs770785559 Chr.7: 4761263-4761263 on Build GRCh38
rs762122739 Chr.7: 4761225-4761225 on Build GRCh38
rs772982227 Chr.7: 4761200-4761200 on Build GRCh38
rs766420272 Chr.7: 4761149-4761149 on Build GRCh38
rs769920475 Chr.7: 4761274-4761274 on Build GRCh38
rs924294173 Chr.7: 4761170-4761170 on Build GRCh38
rs764284651 Chr.7: 4761212-4761212 on Build GRCh38
rs147566143 Chr.7: 4761167-4761167 on Build GRCh38
rs760671379 Chr.7: 4761146-4761146 on Build GRCh38
rs752608579 Chr.7: 4761099-4761099 on Build GRCh38
rs921047378 Chr.7: 4761187-4761187 on Build GRCh38
rs773342241 Chr.7: 4761145-4761145 on Build GRCh38
rs373966273 Chr.7: 4761226-4761226 on Build GRCh38
rs545357409 Chr.7: 4761301-4761301 on Build GRCh38
rs768925427 Chr.7: 4761122-4761122 on Build GRCh38
rs931603387 Chr.7: 4761233-4761233 on Build GRCh38
rs200826976 Chr.7: 4761113-4761113 on Build GRCh38
rs774649096 Chr.7: 4761214-4761214 on Build GRCh38
rs752175698 Chr.7: 4761295-4761295 on Build GRCh38
rs765786255 Chr.7: 4761291-4761291 on Build GRCh38

View Full Product Details