Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 37238393-37238662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCR3
Entrez Gene ID
10311
Gene Name
DSCR3 arrestin fold containing
Gene Aliases
DCRA, DSCRA
UniGene
Hs.369488
Gene Chromosome Location
Chr.21: 37223425-37268124 on Build GRCh38
Transcripts
NM_006052.1 XM_005260909.4 XM_005260911.4 XM_011529423.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7140047 c.259G>T p.G87C Chr.21: 37238552-37238552 on Build GRCh38
260590 c.229G>T p.E77* Chr.21: 37238582-37238582 on Build GRCh38
4654831 c.233T>C p.M78T Chr.21: 37238578-37238578 on Build GRCh38
5715629 c.260G>A p.G87D Chr.21: 37238551-37238551 on Build GRCh38
1632476 c.211A>G p.I71V Chr.21: 37238600-37238600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747618434 Chr.21: 37238552-37238552 on Build GRCh38
rs770503472 Chr.21: 37238436-37238436 on Build GRCh38
rs752773299 Chr.21: 37238498-37238498 on Build GRCh38
rs149254619 Chr.21: 37238566-37238566 on Build GRCh38
rs773451498 Chr.21: 37238482-37238482 on Build GRCh38
rs372016915 Chr.21: 37238578-37238578 on Build GRCh38
rs371871794 Chr.21: 37238455-37238455 on Build GRCh38
rs767754573 Chr.21: 37238497-37238497 on Build GRCh38
rs368525475 Chr.21: 37238439-37238439 on Build GRCh38
rs762332467 Chr.21: 37238448-37238448 on Build GRCh38
rs200696626 Chr.21: 37238435-37238435 on Build GRCh38
rs774113251 Chr.21: 37238437-37238437 on Build GRCh38
rs757143569 Chr.21: 37238538-37238538 on Build GRCh38
rs1028742064 Chr.21: 37238495-37238495 on Build GRCh38
rs746453273 Chr.21: 37238569-37238569 on Build GRCh38
rs765738215 Chr.21: 37238478-37238478 on Build GRCh38
rs777450023 Chr.21: 37238420-37238420 on Build GRCh38
rs776987179 Chr.21: 37238449-37238453 on Build GRCh38
rs201638717 Chr.21: 37238613-37238613 on Build GRCh38
rs766634652 Chr.21: 37238488-37238488 on Build GRCh38
rs112074434 Chr.21: 37238426-37238426 on Build GRCh38
rs771667811 Chr.21: 37238553-37238553 on Build GRCh38
rs773824315 Chr.21: 37238604-37238604 on Build GRCh38
rs1041002211 Chr.21: 37238554-37238554 on Build GRCh38
rs911054616 Chr.21: 37238507-37238507 on Build GRCh38
rs778560708 Chr.21: 37238438-37238438 on Build GRCh38
rs777710237 Chr.21: 37238517-37238517 on Build GRCh38
rs774964036 Chr.21: 37238446-37238446 on Build GRCh38
rs749170688 Chr.21: 37238533-37238533 on Build GRCh38
rs575276995 Chr.21: 37238487-37238487 on Build GRCh38
rs778542605 Chr.21: 37238544-37238544 on Build GRCh38
rs144379747 Chr.21: 37238568-37238568 on Build GRCh38
rs756150340 Chr.21: 37238503-37238503 on Build GRCh38
rs142759447 Chr.21: 37238496-37238496 on Build GRCh38

View Full Product Details