Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38779396-38779647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAL4
Entrez Gene ID
10126
Gene Name
dynein axonemal light chain 4
Gene Aliases
MRMV3, PIG27
UniGene
Hs.632766
Gene Chromosome Location
Chr.22: 38778508-38794156 on Build GRCh38
Transcripts
NM_005740.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3964288 c.250G>A p.E84K Chr.22: 38779517-38779517 on Build GRCh38
221881 c.157G>A p.A53T Chr.22: 38779610-38779610 on Build GRCh38
4104150 c.159C>T p.A53A Chr.22: 38779608-38779608 on Build GRCh38
7263328 c.211G>A p.V71M Chr.22: 38779556-38779556 on Build GRCh38
4398201 c.289C>T p.L97L Chr.22: 38779478-38779478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772899239 Chr.22: 38779624-38779627 on Build GRCh38
rs369007384 Chr.22: 38779556-38779556 on Build GRCh38
rs868115517 Chr.22: 38779453-38779453 on Build GRCh38
rs748198441 Chr.22: 38779487-38779487 on Build GRCh38
rs754739094 Chr.22: 38779571-38779571 on Build GRCh38
rs374991302 Chr.22: 38779461-38779461 on Build GRCh38
rs868430494 Chr.22: 38779584-38779584 on Build GRCh38
rs752638859 Chr.22: 38779578-38779578 on Build GRCh38
rs761381966 Chr.22: 38779547-38779547 on Build GRCh38
rs746167211 Chr.22: 38779620-38779620 on Build GRCh38
rs772575654 Chr.22: 38779539-38779539 on Build GRCh38
rs142229719 Chr.22: 38779448-38779448 on Build GRCh38
rs753342186 Chr.22: 38779425-38779425 on Build GRCh38
rs777878936 Chr.22: 38779602-38779602 on Build GRCh38
rs191545066 Chr.22: 38779433-38779433 on Build GRCh38
rs756201896 Chr.22: 38779599-38779599 on Build GRCh38
rs749569243 Chr.22: 38779514-38779514 on Build GRCh38
rs771262991 Chr.22: 38779515-38779515 on Build GRCh38
rs373776976 Chr.22: 38779611-38779611 on Build GRCh38
rs113358406 Chr.22: 38779545-38779545 on Build GRCh38
rs750166671 Chr.22: 38779434-38779434 on Build GRCh38
rs553838266 Chr.22: 38779518-38779518 on Build GRCh38
rs562794909 Chr.22: 38779544-38779544 on Build GRCh38
rs762739403 Chr.22: 38779550-38779550 on Build GRCh38
rs751354332 Chr.22: 38779557-38779557 on Build GRCh38
rs892161357 Chr.22: 38779546-38779546 on Build GRCh38
rs779128761 Chr.22: 38779613-38779613 on Build GRCh38
rs759923818 Chr.22: 38779521-38779521 on Build GRCh38
rs544389269 Chr.22: 38779467-38779467 on Build GRCh38
rs1000901487 Chr.22: 38779472-38779472 on Build GRCh38
rs1029903676 Chr.22: 38779566-38779566 on Build GRCh38
rs749481067 Chr.22: 38779610-38779610 on Build GRCh38
rs142612396 Chr.22: 38779572-38779572 on Build GRCh38
rs558208634 Chr.22: 38779430-38779430 on Build GRCh38
rs746629700 Chr.22: 38779490-38779490 on Build GRCh38
rs112106258 Chr.22: 38779628-38779628 on Build GRCh38
rs769764294 Chr.22: 38779492-38779492 on Build GRCh38
rs912710005 Chr.22: 38779418-38779418 on Build GRCh38
rs764712598 Chr.22: 38779548-38779548 on Build GRCh38
rs771733261 Chr.22: 38779421-38779422 on Build GRCh38
rs754907328 Chr.22: 38779466-38779466 on Build GRCh38
rs758258970 Chr.22: 38779437-38779437 on Build GRCh38
rs778048428 Chr.22: 38779506-38779506 on Build GRCh38

View Full Product Details